Skip to Content
Merck
All Photos(9)

Key Documents

HPA001922

Sigma-Aldrich

Anti-RAP1GAP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Rap1 GTPase-activating protein 1 antibody produced in rabbit, Anti-Rap1GAP antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

IGIENIQEVQEKRESPPAGQKTPDSGHVSQEPKSENSSTQSSPEMPTTKNRAETAAQRAEALKDFSRSSSSASSFASVVEETEGVDGEDTGLESVSSSGTP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAP1GAP(5909)

General description

RAP1GAP (RAP1 GTPase activating protein) is a Ras-like guanine-nucleotide-binding protein (GNBP), expressed abundantly in fetal tissues and certain tumor cell lines, such as Wilms′ kidney tumor, SK-NEP-1, and the melanoma. Its expression is mainly in brain and to lesser extent in the lymphoid tissues. In addition to unique functional domains, it also contains GAP-related domain (GRD), responsible for GAP catalytic activity.

Immunogen

Rap1 GTPase-activating protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RAP1GAP (RAP1 GTPase activating protein) is involved with various signal transduction pathways such as integrin-mediated cell adhesion, cell proliferation, differentiation, activation of extracellular signal-regulated kinase. The activity of Rap1 is regulated by guanine-nucleotide-exchange factors (GEFs) and GTPase-activating proteins (GAPs) along with other extracellular stimuli.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86386

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Daumke et al.
Nature, 429(6988), 197-201 (2004-05-14)
Rap1 is a Ras-like guanine-nucleotide-binding protein (GNBP) that is involved in a variety of signal-transduction processes. It regulates integrin-mediated cell adhesion and might activate extracellular signal-regulated kinase. Like other Ras-like GNBPs, Rap1 is regulated by guanine-nucleotide-exchange factors (GEFs) and GTPase-activating
B Rubinfeld et al.
Cell, 65(6), 1033-1042 (1991-06-14)
The rap1/Krev-1 gene encodes a ras-related protein that suppresses transformation by ras oncogenes. We have purified an 88 kd GTPase activating protein (GAP), specific for the rap1/Krev-1 gene product, from bovine brain. Based on partial amino acid sequences obtained from
Li Su et al.
The Journal of biological chemistry, 278(17), 15232-15238 (2003-02-19)
In the present study, we showed that SPA-1, a Rap1 GTPase-activating protein (GAP), was bound to a cytoskeleton-anchoring protein AF-6. SPA-1 and AF-6 were co-immunoprecipitated in the 293T cells transfected with both cDNAs as well as in normal thymocytes. In
H Kurachi et al.
The Journal of biological chemistry, 272(44), 28081-28088 (1997-11-05)
Mouse Spa-1 gene with a region homologous to the human rap1GAP gene is transcriptionally induced in the lymphocytes by mitogenic stimulation. Herein we have cloned a cDNA for its human counterpart. SPA-1 cDNA encodes a 130-kDa protein (p130(SPA-1)) consisting of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service