Skip to Content
Merck
All Photos(6)

Key Documents

HPA000848

Sigma-Aldrich

Anti-FBLN5 antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dance antibody produced in rabbit, Anti-Developmental arteries and neural crest EGF-like protein antibody produced in rabbit, Anti-FIBL-5 antibody produced in rabbit, Anti-Fibulin-5 precursor antibody produced in rabbit, Anti-UP50 antibody produced in rabbit, Anti-Urine p50 protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

TYFCSCPPGYILLDDNRSCQDINECEHRNHTCNLQQTCYNLQGGFKCIDPIRCEEPYLRISDNRCMCPAENPGCRDQPFTILYRDMDVVSGRSVPADIFQMQATTRYPGAYYIFQIKSGNEGREFYMRQTGPISATLVMTRPIKGPR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBLN5(10516)

Looking for similar products? Visit Product Comparison Guide

General description

Fibulin-5 is a protein encoded by the FBLN5 gene in humans and is mapped to chromosome 14q31.

Immunogen

Fibulin-5 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Fibulin-5 is a secreted extracellular matrix glycoprotein that has diverse panel of biological functions. It can segregate into elastogenic as well as extra-elastogenic functions. FBLN5 mediates cell adherence through subset of integrins, antagonizes angiogenesis and inhibits migration as well as proliferation of endothelial and smooth muscle cells. The gene is associated with primary spontaneous pneumothorax (PSP) in young PSP patients. Higher expression of FBLN5 contributes to the development of new therapeutic strategies in the FLJ10540/AKT pathway for Nasopharyngeal carcinoma (NPC) treatment. The gene is closely related to carcinoma metastasis and may play a negative role in the development as well as metastasis of ovarian cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70454

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abigail Kate Langton et al.
The Journal of investigative dermatology, 139(4), 779-788 (2018-11-08)
Skin aging is a complex process involving the additive effects of time-dependent intrinsic aging and changes elicited via skin's interaction with the environment. Maintaining optimal skin function is essential for healthy aging across global populations; yet most research focuses on
Abigail Kate Langton et al.
The Journal of investigative dermatology, 139(4), 779-788 (2018-11-08)
Skin aging is a complex process involving the additive effects of time-dependent intrinsic aging and changes elicited via skin's interaction with the environment. Maintaining optimal skin function is essential for healthy aging across global populations; yet most research focuses on
A K Langton et al.
The British journal of dermatology, 171(2), 274-282 (2014-02-04)
Geographical ancestry plays a key role in determining the susceptibility of human skin to external insults and dermatological disease. Despite this, studies of skin from individuals of diverse geographical ancestry focus primarily on epidermal pigmentation. Few reports characterize the gross
A K Langton et al.
The British journal of dermatology, 177(3), 818-827 (2017-01-31)
Human skin has the crucial roles of maintaining homeostasis and protecting against the external environment. Skin offers protection against mechanical trauma due to the reversible deformation of its structure; these biomechanical properties are amenable to dynamic testing using noninvasive devices.
Amy Louise Winship et al.
Oncology letters, 12(1), 651-657 (2016-06-28)
Endometrial cancer is the most common invasive gynaecological malignancy. While endocrine, genetic and inflammatory factors are thought to contribute to its pathogenesis, its precise etiology and molecular regulators remain poorly understood. Fibulin-5 is an extracellular matrix (ECM) protein that inhibits

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service