Skip to Content
Merck
All Photos(1)

Documents

H6666

Sigma-Aldrich

Keratinocyte Growth Factor human

HumanKine®, >95% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

Synonym(s):

FGF-7, KGF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Keratinocyte Growth Factor human, KGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

4-20 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 17-30 kDa (glycosylated)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGF7(2252)

Looking for similar products? Visit Product Comparison Guide

General description

Keratinocyte Growth Factor (KGF) belongs to the fibroblast growth factor 7 (FGF-7) family. It is produced by the mesenchymal cells, γδ T cells, and fibroblasts in several organs. The KGF gene is located on the human chromosome at 15q21.2.

Application

Keratinocyte Growth Factor (KGF) human has been used as a component of the keratinocyte differentiation medium.

Biochem/physiol Actions

Keratinocyte Growth Factor (KGF) exerts paracrine effects on mesenchymal cells. It exhibits cryoprotective effects on epithelial cells of the lungs, gastrointestinal tract, and the lungs. KGF participates in the activation of IIIb isoform of the fibroblast growth factor (FGF)-receptor 2 (FGFR2-IIIb) present on keratinocytes. This activation further favors proliferation, migration, and survival of keratinocytes.

Physical form

Lyophilized from a 0.2 μm filtered solution of 1x PBS.

Preparation Note

HumanKine Keratinocyte Growth Factor (KGF) is expressed as glycosylated 17 and 30 kDa monomers in human HEK 293 cells. Production in human HEK 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The activity was determined by the dose-dependent stimulation of the proliferation of 4MBr-5 cells (monkey epithelial cell line).

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qandeel Fatima et al.
Open life sciences, 13, 544-552 (2018-12-31)
Autologous skin grafts are used to treat severe burn wounds, however, the availability of adequate donor sites makes this option less practical. Recently, stem cells have been used successfully in tissue engineering and in regenerative medicine. The current study aims
Michael J LaFemina et al.
American journal of physiology. Lung cellular and molecular physiology, 299(6), L724-L734 (2010-09-14)
Keratinocyte growth factor (KGF) has efficacy in several experimental models of lung injury; however, the mechanisms underlying KGF's protective effect remain incompletely understood. This study was undertaken to determine whether KGF augments barrier function in primary rat alveolar epithelial cells
Verônica Chomiski et al.
Acta cirurgica brasileira, 31(8), 505-512 (2016-09-01)
To evaluate the effect of keratinocyte growth factor (KGF) treatment on the expression of wound-healing-related genes in cultured keratinocytes from burn patients. Keratinocytes were cultured and divided into 4 groups (n=4 in each group): TKB (KGF-treated keratinocytes from burn patients)
Umbilical cord tissue derived mesenchymal stem cells can differentiate into skin cells
Fatima Q, et al.
The Open Immunology Journal, 13(1), 544-552 (2018)
Susanne Braun et al.
Journal of cell science, 119(Pt 23), 4841-4849 (2006-11-09)
Owing to its potent cytoprotective properties for epithelial cells, keratinocyte growth factor (KGF) is successfully used for the treatment of chemotherapy- and radiotherapy-induced oral mucositis in cancer patients. It is therefore of major interest to determine possible clinical applications of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service