Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1342-01

ExcelGelSDS buffer strips

L × W 245 mm × 4.5 mm, pack of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

Pricing and availability is not currently available.

description

Polyacrylamide T = 12%, C = 3%

feature

wetted part: no

packaging

pack of 1 kit

manufacturer/tradename

Cytiva 17134201

L × W

245 mm × 4.5 mm

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GE80-1261-01MPMESMPM0PS
packaging

pack of 1 kit

packaging

pack of 6 ea

packaging

pack of 5 ea (Each packet makes 1 liter of running buffer)

packaging

pack of 5 ea (Each packet makes 1 liter of running buffer)

description

Polyacrylamide T = 12%, C = 3%

description

-

description

-

description

-

feature

wetted part: no

feature

wetted part: no

feature

-

feature

-

L × W

245 mm × 4.5 mm

L × W

-

L × W

-

L × W

-

General description

ExcelGel SDS Buffer Strips (anode and cathode)

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

ExcelGel is a trademark of Cytiva

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service