Skip to Content
Merck
All Photos(2)

Key Documents

G6160

Sigma-Aldrich

Monoclonal Anti-β-COP antibody produced in mouse

clone maD, ascites fluid

Synonym(s):

Anti-BARMACS, Anti-COPB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

maD, monoclonal

contains

15 mM sodium azide

species reactivity

human, rat, hamster, monkey

technique(s)

indirect immunofluorescence: 1:80 using cultured Chinese hamster ovary (CHO) cells
microarray: suitable
western blot: 1:1,000 using a preparation of stacked Golgi membranes from rat liver

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COPB1(1315)
rat ... Copb1(114023)

General description

The coatomer (approx. 550kDa) consists of proteins designated α-, β-, γ-, and δ-COP, together with substoichiometric amounts of several other proteins.

Specificity

The antibody recognizes an epitope in the β-COP protein (110 kDa) and stains the periphery of the Golgi complex using immunocytochemical techniques.

Immunogen

synthetic peptide D1 of β-COP (a.a. 701-715) conjugated to KLH

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Monoclonal Anti-β-COP antibody produced in mouse is suitable for use as a primary antibody in immunoblot:
  • analysis at a working dilution of 1:1000 using subcellular proteins from rat PC12 (pheochromocytoma) cells
  • analysis of gradient fractions of cerebral microvessels to confirm the separation of plasma membrane lipid raft domains from intracellular membranous components
  • detection of the Golgi marker protein β-COP in exosome-enriched extracellular microvesicles (eMV) preparations from untreated HeLa cells

It is suitable for immunostaining of β-coatomer, that is used as a intracellular, Golgi protein marker :
  • to confirm that CD14 staining is localized to the cell surface of HAEC
  • for examining the localization of Meltrin β in the Golgi apparatus and its vicinity in neurons prepared from developing dorsal root ganglia of mouse embryos
  • in NB4 and NB4-LR1 cells to examine the colocalization of PKA regulatory subunits
  • in CHO cells to examine colocalization of GFP-Rab24

It is suitable for use in cell-surface ELISA of human aortic endothelial cells (HAEC)

It is also suitable for western blot analysis at a working dilution of 1:1000 using a preparation of stacked Golgi membranes from rat liver, for indirect immunofluorescence at a working dilution of 1:80 using cultured Chinese hamster ovary (CHO) cells and for microarray.

Biochem/physiol Actions

COPs (coatomer proteins) contain adaptin-like, complex (8) and are transiently attached to the vesicles involved in transport within the Golgi complex and possibly between the rough ER and Golgi complex. β-COP has a molar mass of 110kDa and its primary structure is homologous to the β-adaptin component of clathrin-coated vesicles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumi Kano et al.
Journal of cell science, 122(Pt 13), 2218-2227 (2009-06-11)
Yip1A, a mammalian homologue of yeast Yip1p, is a multi-spanning membrane protein that is considered to be involved in transport between the endoplasmic reticulum (ER) and the Golgi. However, the precise role of Yip1A in mammalian cells remains unclear. We
J W Creemers et al.
The Journal of biological chemistry, 276(6), 4211-4217 (2000-11-10)
The amyloid peptide is the main constituent of the amyloid plaques in brain of Alzheimer's disease patients. This peptide is generated from the amyloid precursor protein by two consecutive cleavages. Cleavage at the N terminus is performed by the recently
Daniela B Munafó et al.
Traffic (Copenhagen, Denmark), 3(7), 472-482 (2002-06-06)
Rab GTPases comprises a large family of proteins, with more than 50 gene products localized in distinct subcellular compartments. Rab24 is a member of this family whose function is not presently known. In order to elucidate the role of this
A Oprins et al.
The Journal of cell biology, 121(1), 49-59 (1993-04-01)
We examined the distribution of the non-clathrin-coated vesicle-associated coat protein beta-COP in rat exocrine pancreatic cells by immunogold cytochemistry. Labeling for beta-COP was found in the Golgi region (48%) where it was associated with vesicles and buds of approximately 50
Eric Nguyen et al.
Molecular pharmacology, 83(5), 1057-1065 (2013-03-05)
Acute promyelocytic leukemia (APL) is characterized by granulopoietic differentiation arrest at the promyelocytic stage. In most cases, this defect can be overcome by treatment with all-trans-retinoic acid (ATRA), leading to complete clinical remission. Cyclic AMP signaling has a key role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service