Skip to Content
Merck
All Photos(1)

Key Documents

EP0030122348

Eppendorf® DNA LoBind tubes

capacity 5 mL, cap (screw cap), PCR clean, case of 200 ea (2 bags of 100 Tubes each)

Synonym(s):

Eppendorf® DNA LoBind, Eppendorf Tube®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

feature

PCR clean
cap (screw cap)

packaging

case of 200 ea (2 bags of 100 Tubes each)

manufacturer/tradename

Eppendorf® 0030122348

capacity

5 mL

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf LoBind® Tubes are made from a special polymer and the LoBind material helps to enhance the recovery rate of samples. These single-use tubes, made from polypropylene, are ideal for a wide range of applications such as sample preparation, mixing, centrifugation, transportation, and storage of solid and liquid samples and reagents. Eppendorf LoBind® Tubes are available in two product versions:
  • Eppendorf Protein LoBind is specifically designed for the preparation and storage of protein, peptide, antibody, or virus samples.
  • DNA LoBind is suitable for DNA or RNA applications in forensic analysis, microarrays, and next-generation sequencing.

Application

Eppendorf® DNA LoBind, Eppendorf Tube® has been used for cDNA purification using magnetic beads.

Features and Benefits

  • Designed to meet rigorous demands for centrifugation, temperature resistance, and chemical compatibility.
  • Equipped with a secure screw cap.
  • Designed with a graduation feature to indicate the filling quantity, along with a labeling area for easy sample identification.
  • The tubes are certified to be human DNA, DNase, RNase, and PCR inhibitors (PCR clean) free.
Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf LoBind is a registered trademark of Eppendorf AG
Eppendorf Tube is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik S Schild et al.
STAR protocols, 2(2), 100411-100411 (2021-04-20)
RNA tomography or tomo-seq combines mRNA sequencing and cryo-sectioning to spatially resolve gene expression. We have adapted this method for the nematode Caenorhabditis elegans to generate anteroposterior gene expression maps at near-cellular resolution. Here, we provide a detailed overview of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service