Skip to Content
Merck
All Photos(1)

Key Documents

EHU088381

Sigma-Aldrich

MISSION® esiRNA

targeting human EGLN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCTGGCGCTCGAGTACATCGTGCCGTGCATGAACAAGCACGGCATCTGTGTGGTGGACGACTTCCTCGGCAAGGAGACCGGACAGCAGATCGGCGACGAGGTGCGCGCCCTGCACGACACCGGGAAGTTCACGGACGGGCAGCTGGTCAGCCAGAAGAGTGACTCGTCCAAGGACATCCGAGGCGATAAGATCACCTGGATCGAGGGCAAGGAGCCCGGCTGCGAAACCATTGGGCTGCTCATGAGCAGCATGGACGACCTGATACGCCACTGTAACGGGAAGCTGGGCAGCTACAAAATCAATGGCCGGACGAAAGCCATGGTTGCTTGTTATCCGGGCAATGGAACGGGTTATGTACGTCATGTTGATAATCCAAATGGAGATGGAAGATGTGTGAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junqiang Guo et al.
Artificial cells, nanomedicine, and biotechnology, 48(1), 37-45 (2019-12-20)
Background: Prolyl hydroxylase domain proteins (PHD2) is an oxygen sensor that is able to induce hypoxia-inducible factor-α (HIF-α) degradation under normoxic condition. The present paper designed to reveal the function of PHD2 in hepatocellular carcinoma (HCC) cells proliferation, migration and
Hyo Jeong Yong et al.
Oncotarget, 8(41), 69833-69846 (2017-10-21)
Hypoxia-induced interleukin-32β (IL-32β) shifts the metabolic program to the enhanced glycolytic pathway. In the present study, the underlying mechanism by which hypoxia-induced IL-32β stability is regulated was investigated in ovarian cancer cells. IL-32β expression increased under hypoxic conditions in ovarian
Junping Hu et al.
Scientific reports, 7(1), 15878-15878 (2017-11-22)
Proteinuria is closely associated with the progression of chronic kidney diseases (CKD) by producing renal tubulointerstitial fibrosis. Over-activation of hypoxia inducible factor (HIF)-1α has been implicated in the progression of CKD. The present study tested the hypothesis that HIF-1α mediates
Anindya Dey et al.
Science advances, 6(27) (2020-09-17)
The stringent expression of the hypoxia inducible factor-1α (HIF-1α) is critical to a variety of pathophysiological conditions. We reveal that, in normoxia, enzymatic action of cystathionine β-synthase (CBS) produces H2S, which persulfidates prolyl hydroxylase 2 (PHD2) at residues Cys21 and
Kai Zhu et al.
International journal of nanomedicine, 9, 5203-5215 (2014-11-28)
Mesenchymal stem cell (MSC) transplantation has attracted much attention in myocardial infarction therapy. One of the limitations is the poor survival of grafted cells in the ischemic microenvironment. Small interfering RNA-mediated prolyl hydroxylase domain protein 2 (PHD2) silencing in MSCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service