Skip to Content
Merck
All Photos(1)

Key Documents

EHU061741

Sigma-Aldrich

MISSION® esiRNA

targeting human BECN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCTGAGAGACTGGATCAGGAGGAAGCTCAGTATCAGAGAGAATACAGTGAATTTAAACGACAGCAGCTGGAGCTGGATGATGAGCTGAAGAGTGTTGAAAACCAGATGCGTTATGCCCAGACGCAGCTGGATAAGCTGAAGAAAACCAACGTCTTTAATGCAACCTTCCACATCTGGCACAGTGGACAGTTTGGCACAATCAATAACTTCAGGCTGGGTCGCCTGCCCAGTGTTCCCGTGGAATGGAATGAGATTAATGCTGCTTGGGGCCAGACTGTGTTGCTGCTCCATGCTCTGGCCAATAAGATGGGTCTGAAATTTCAGAGATACCGACTTGTTCCTTACGGAAACCATTCATATCTGGAGTCTCTGACAGACAAATCTAAGGAGCTGCCGTTATACTGTTCTGGGGGGTTGCGGTTTTTCTGGGACAACAAGTTTGACCATGCAATGGTGGCTTTCCTGGACTGTGTGCAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingjing Wu et al.
Journal of cellular and molecular medicine, 22(2), 1190-1201 (2017-10-28)
Long-term peritoneal dialysis is accompanied by functional and histopathological alterations in the peritoneal membrane. In the long process of peritoneal dialysis, high-glucose peritoneal dialysis solution (HGPDS) will aggravate the peritoneal fibrosis, leading to decreased effectiveness of peritoneal dialysis and ultrafiltration
Haoran Peng et al.
Viruses, 10(5) (2018-05-16)
Autophagy is a common strategy for cell protection; however, some viruses can in turn adopt cellular autophagy to promote viral replication. Zika virus (ZIKV) is the pathogen that causes Zika viral disease, and it is a mosquito-borne virus. However, its
Michelle M Coleman et al.
American journal of respiratory cell and molecular biology, 59(5), 548-556 (2018-06-01)
Vitamin A deficiency strongly predicts the risk of developing tuberculosis (TB) in individuals exposed to Mycobacterium tuberculosis (Mtb). The burden of antibiotic-resistant TB is increasing globally; therefore, there is an urgent need to develop host-directed adjunctive therapies to treat TB.
Wenkang Luan et al.
OncoTargets and therapy, 10, 4569-4577 (2017-10-27)
Autophagy is not only a survival response to growth-factor or nutrient deprivation but also an important mechanism for tumor-cell suicide, including melanoma.
Guangmin Xi et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 1610-1616 (2016-11-09)
Multidrug resistance (MDR) is a major obstacle for successful chemotherapy treatment. Searching for effective MDR modulators and combining them with anticancer drug therapies has been a promising strategy against clinical MDR. In our previous study, we have found that DHA-E3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service