Skip to Content
Merck
All Photos(1)

Documents

C5624

Sigma-Aldrich

Cryptotanshinone

≥98% (HPLC)

Synonym(s):

1,2,6,7,8,9-hexahydro-1,6,6-trimethyl- (R)-phenanthro(1,2-b)furan-10,11-dione, Cryptotanshinon, Tanshinone c

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20O3
CAS Number:
Molecular Weight:
296.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -80 to -99°, c = 0.5 in chloroform-d

storage condition

desiccated
protect from light

color

orange-brown

solubility

DMSO: ≥5 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C[C@H]1COC2=C1C(=O)C(=O)c3c4CCCC(C)(C)c4ccc23

InChI

1S/C19H20O3/c1-10-9-22-18-12-6-7-13-11(5-4-8-19(13,2)3)15(12)17(21)16(20)14(10)18/h6-7,10H,4-5,8-9H2,1-3H3/t10-/m0/s1

InChI key

GVKKJJOMQCNPGB-JTQLQIEISA-N

Biochem/physiol Actions

Cryptotanshinone is a quinoid diterpene isolated from the root of the Asian medicinal plant, Salvia miotiorrhiza bunge. Recently it was discovered that the compound is a potent STAT3 inhibitor. Cryptotanshinone rapidly inhibited STAT3 Tyr705 phosphorylation through a JAK2-independent mechanism. Cryptotanshinone selectively inhibits STAT3-activated cell lines through binding to monomer STAT3, subsequently blocking the dimerization and inhibiting STAT3 transcriptional regulatory activity. Previously, it was reported that the compound counteracts inflammation through the inhibition of cyclooxygenase II activity and endothelin-1 expression. In traditional oriental medicine dried roots of Salvia Miltiorrhiza Bunge (Danshen) have commonly been used for the treatment of circulatory disorders, liver disease, coronary heart disease, hepatitis, and chronic renal failure.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Hau Yen et al.
Molecular therapy oncolytics, 26, 158-174 (2022-07-22)
Dampening tumor growth by converting tumor-associated macrophages (TAMs) from M2/repair-types to M1/kill-types is of high interest. Here, we show that cryptotanshinone (CPT) can function as an antitumor immune modulator that switches TAMs from an M2 to an M1 phenotype, leading
Zhiyuan Lu et al.
Molecular biology reports, 51(1), 436-436 (2024-03-23)
Elevated levels of adipokine chemerin have been identified in oral squamous cell carcinoma (OSCC) and found to be associated with metastasis to the cervical lymph nodes. The underlying mechanism through which chemerin affects OSCC progression is unclear. The aims of
Yongpeng Zhang et al.
Drug development research, 82(4), 581-588 (2020-12-20)
Coronary microembolization (CME) is a prevalent cardiovascular disease, especially nowadays when percutaneous coronary intervention is widely applied. However, neither cardio-protective agents nor devices for distal protection could effectively prevent the occurrence of CME. Therefore, we aimed to develop a new
Defeng Xu et al.
The Prostate, 72(7), 803-816 (2011-09-21)
Androgen receptor (AR) is the main therapeutic target for the treatment of prostate cancer (PCa). Anti-androgens to reduce or prevent androgens binding to AR are widely used to suppress AR-mediated PCa growth; however, the androgen depletion therapy (ADT) is only
Yan Luo et al.
Cancer prevention research (Philadelphia, Pa.), 4(12), 2083-2091 (2011-09-02)
Cryptotanshinone (CPT), isolated from the plant Salvia miltiorrhiza Bunge, is a potential anticancer agent. However, the underlying mechanism remains to be defined. Here, we show that CPT inhibited lymphangiogenesis in an in vitro model (tube formation). This effect was partly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service