Skip to Content
Merck
All Photos(1)

Documents

C2997

Sigma-Aldrich

Cytosporone B

≥98% (HPLC), powder, nuclear receptor Nur77 agonist

Synonym(s):

3,5-Dihydroxy-2-(1-oxooctyl)benzeneacetic acid ethyl ester, Csn-B, Csn-B; 3,5-Dihydroxy-2-(1-oxooctyl)-benzeneacetic acid, ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O5
CAS Number:
Molecular Weight:
322.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Cytosporone B, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

−20°C

SMILES string

CCCCCCCC(=O)c1c(O)cc(O)cc1CC(=O)OCC

InChI

1S/C18H26O5/c1-3-5-6-7-8-9-15(20)18-13(11-17(22)23-4-2)10-14(19)12-16(18)21/h10,12,19,21H,3-9,11H2,1-2H3

InChI key

UVVWQQKSNZLUQA-UHFFFAOYSA-N

Application

Cytosporone B has been used:
  • as a positive control and NR4A1 agonist in NR4A1 reporter gene assay
  • for Nr4a1 activation
  • to treat OVX mice and study its effect on migration of osteoclast precursor

Biochem/physiol Actions

Cytosporone B (Csn-B) is the first naturally occurring agonist for nuclear orphan receptor Nur77. It binds with high affinity (IC50=0.278 nM) to the ligand-binding domain of Nur77 and stimulates Nur77-dependent activities.

Nur77 is a nuclear receptor/transcription factor. A physiological ligand for Nur77 is as yet unknown, but there is increasing interest in Nur77 because of its known activities. Translocation of Nur77 from the nucleus to mitochondria initiates cell apoptosis, making it a potential target for cancer treatment. Nur77 is also involved in glucose homeostasis; it induces genes involved in gluconeogenesis. Csn-B physically binds to Nur77 and activates its transactivational activity and translocation to mitochondria to induce apoptosis. It inhibits cancer cell proliferation and tumor growth.
Cytosporone B is a fungal metabolite closely related to phomposin C. It is the first known agonist for the nuclear orphan receptor Nur77. It binds with high affinity (IC50 = 0.278 nM) to the ligand-binding domain of Nur77 and stimulates Nur77-dependent activities.

Nur77 is a nuclear receptor/transcription factor with no known physiological ligand, but there is increasing interest in Nur77 because of its known activities. Translocation of Nur77 from the nucleus to mitochondria initiates apoptosis, making it a potential target for cancer chemotherapy. Nur77 also induces genes involved in gluconeogenesis. Csn-B activates the Nur77 translocation to mitochondria to induce apoptosis, inhibiting cancer cell proliferation and tumor growth.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aravind T Reddy et al.
PloS one, 15(2), e0229256-e0229256 (2020-02-23)
Cigarette smoke (CS) contains multiple gaseous and particulate materials that can cause lung inflammation, and smoking is the major cause of chronic obstructive pulmonary disease (COPD). We sought to determine the mechanisms of how CS triggers lung inflammation. Nur77, a
Mohammad Ishaq et al.
Innate immunity, 26(8), 746-758 (2020-09-15)
Nuclear hormone receptor ligands are known to modulate innate immunity by dampening the immune response induced by pathogens. Here, we report that unlike other ligands, 3,3',5-triiodo-l-thyronine (T3) induced the type 1 IFN response and expression of IFN-stimulated genes (ISGs). T3
Pro-angiogenic Ginsenosides F1 and Rh1 Inhibit Vascular Leakage by Modulating NR4A1
Kang JI, et al.
Scientific reports, 9(1), 4502-4502 (2019)
Benoit Egarnes et al.
PloS one, 12(10), e0186639-e0186639 (2017-10-21)
The transcription factor NR4A1 has emerged as a pivotal regulator of the inflammatory response and immune homeostasis. Although contribution of NR4A1 in the innate immune response has been demonstrated, its role in host defense against viral infection remains to be
Shirley Dehn et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 32(1), 254-264 (2017-09-02)
Phagocytosis after myocardial infarction (MI) is a prerequisite to cardiac repair. Recruited monocytes clear necrotic cardiomyocytes and differentiate into cardiac macrophages. Some studies have linked apoptotic cell receptors on cardiac macrophages to tissue repair; however, the contribution of precursor monocyte

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service