Skip to Content
Merck
All Photos(1)

Key Documents

C197

Sigma-Aldrich

8-(3-Chlorostyryl)caffeine

≥98% (HPLC), solid

Synonym(s):

1,3,7-Trimethyl-8-(3-chlorostyryl)xanthine, CSC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15ClN4O2
CAS Number:
Molecular Weight:
330.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >5 mg/mL

storage temp.

−20°C

SMILES string

[H]\C(=C(\[H])c1nc2C(=O)N(C)C(=O)N(C)c2n1C)c3cccc(Cl)c3

InChI

1S/C16H15ClN4O2/c1-19-12(8-7-10-5-4-6-11(17)9-10)18-13-14(19)20(2)16(23)21(3)15(13)22/h4-9H,1-3H3/b8-7+

InChI key

MHYRUZOJQQLLQS-BQYQJAHWSA-N

Gene Information

Application

8-(3-Chlorostyryl)caffeine has been used as an adenosine A2A receptor antagonist:
  • to study its effects on levodopa (L-DOPA)-induced dopamine (DA) release in rat striatum
  • to study its effect on the nuclear factor κ-light-chain-enhancer of activated B cells (NF-κB) activity in macrophages
  • to determine its effects on human umbilical vein endothelial cells (HUVECs)

Biochem/physiol Actions

8-(3-Chlorostyryl)caffeine is a selective adenosine A2A receptor antagonist and monoamine oxidase B (MAO B) inhibitor.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Photosensitive, store in the dark

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krystyna Gołembiowska et al.
Brain research, 998(2), 208-217 (2004-01-31)
In the present study, we investigated effects of the new selective adenosine A2A receptor antagonist 8-(3-chlorostyryl)caffeine (CSC) on L-DOPA-induced dopamine (DA) release in the striatum of intact and reserpine-treated rats. CSC given in a pharmacologically effective dose of 5 mg/kg
Rangel L Silva et al.
Toxicology and applied pharmacology, 368, 63-71 (2019-02-24)
Cannabidiol (CBD) is a natural compound with psychoactive therapeutic properties well described. Conversely, the immunological effects of CBD are still poorly explored. In this study, the potential anti-inflammatory effects and underlying mechanisms of CBD and its analog Dimethyl-Heptyl-Cannabidiol (DMH-CBD) were
K A Jacobson et al.
FEBS letters, 323(1-2), 141-144 (1993-05-24)
An adenosine antagonist, 8-(3-chlorostyryl)caffeine (CSC), was shown previously to be 520-fold selective for A2a-adenosine receptors in radioligand binding assays in the rat brain. In reversing agonist effects on adenylate cyclase, CSC was 22-fold selective for A2a receptors in rat phenochromocytoma
K A Jacobson et al.
Journal of medicinal chemistry, 36(10), 1333-1342 (1993-05-14)
A series of substituted 8-styryl derivatives of 1,3,7-alkylxanthines was synthesized as potential A2-selective adenosine receptor antagonists, and the potency at rat brain A1- and A2-receptors was studied in radioligand binding experiments. At the xanthine 7-position, only small hydrophobic substituents were
Eduardo P Amaral et al.
The Journal of infectious diseases, 219(6), 964-974 (2018-10-12)
Tuberculous pneumonia, necrotic granulomatous lesions, and bacterial dissemination characterize severe forms of mycobacterial infection. To evaluate the pulmonary CD4+ T-cell response during severe tuberculosis, C57BL/6 mice were infected with approximately 100 bacilli of 3 hypervirulent mycobacterial isolates (Mycobacterium tuberculosis strain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service