Skip to Content
Merck
All Photos(1)

Key Documents

A9361

Sigma-Aldrich

Artemether

≥98% (HPLC)

Synonym(s):

Dihydroartemisinin methyl ether, Dihydroqinghaosu methyl ether, SM-224

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26O5
CAS Number:
Molecular Weight:
298.37
MDL number:
UNSPSC Code:
51101908
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +155 to +175°, c = 0.5 in methanol

color

off-white to light brown

solubility

DMSO: ≥20 mg/mL

originator

Novartis

storage temp.

room temp

SMILES string

CO[C@H]1OC2O[C@@]3(C)CCC4[C@H](C)CCC([C@H]1C)[C@@]24OO3

InChI

1S/C16H26O5/c1-9-5-6-12-10(2)13(17-4)18-14-16(12)11(9)7-8-15(3,19-14)20-21-16/h9-14H,5-8H2,1-4H3/t9-,10-,11+,12+,13+,14-,15-,16-/m1/s1

InChI key

SXYIRMFQILZOAM-HVNFFKDJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Artemisinin (ART) is a natural compound present in Artemisia annua, a traditional Chinese plant.

Application

Artemether has been used:
  • as an anti-schistosomal compound to test it effect on the larval stages of S. mansoni
  • to sensitize mouse embryonic fibroblasts (MEFs) and human osteosarcoma HT1080 cells to cysteine starvation (STV)-induced ferroptosis
  • to stimulate islets and its effect on α to β transdifferentiation

Biochem/physiol Actions

Artemether is a methyl ether derivative of artemisinin. It is used against multi-drug resistant strains of the malaria parasite, Plasmodium falciparum, and shows potential in treatment of schistosomiasis.
Artemisinin possesses a highly reactive endoperoxide bridge, which is core for its therapeutic potential. The endoperoxide bond reacts with iron in the erythrocytes with malarial parasite. This leads to the generation of reactive oxygen species (ROS) directly targeting the parasite. Artemisinin also regulates ferroptosis in tumor cells. The α cell transcription factor Arx expression is reduced by artemether. Prolonged exposure of primary islets also resulted in loss if identity in endocrine cell types and their functionality.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Org. Perox. D

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sören Frahm et al.
PLoS neglected tropical diseases, 13(1), e0006590-e0006590 (2019-01-29)
The arsenal in anthelminthic treatment against schistosomiasis is limited and relies almost exclusively on a single drug, praziquantel (PZQ). Thus, resistance to PZQ could constitute a major threat. Even though PZQ is potent in killing adult worms, its activity against
Shuhua Xiao et al.
Acta tropica, 82(2), 175-181 (2002-05-22)
Two decades ago, a group of Chinese scientists discovered the antischistosomal properties of artemether, a derivative of the antimalarial drug artemisinin. However, it was only recently that the importance of this finding was recognized internationally, following a collaborative effort between
J Utzinger et al.
Current medicinal chemistry, 8(15), 1841-1860 (2002-01-05)
Human schistosomiasis, a chronic and debilitating parasitic disease of the tropics, is ranked second after malaria in terms of public health importance. At present, there is no vaccine available, and chemotherapy is the cornerstone of schistosomiasis control. Praziquantel is the
Dominic Mosha et al.
Antimicrobial agents and chemotherapy, 58(8), 4583-4592 (2014-05-29)
Artemether-lumefantrine (AL) is the first-line treatment for uncomplicated malaria in the second and third trimesters of pregnancy. Its efficacy during pregnancy has recently been challenged due to altered pharmacokinetic (PK) properties in this vulnerable group. The aim of this study
Abel Nhama et al.
Malaria journal, 13, 309-309 (2014-08-12)
Mozambique adopted artemisinin-based combination therapy (ACT) for the treatment of uncomplicated Plasmodium falciparum malaria in the year 2006, and since 2009 artemether-lumefantrine (AL) and artesunate-amodiaquine (ASAQ) have been proposed as alternative first-line treatments. A multicentre study was conducted in five

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service