Skip to Content
Merck
All Photos(2)

Documents

A5512

Sigma-Aldrich

Aristolochic acid I

≥90% (HPLC), powder, phospholipase A₂ inhibitor

Synonym(s):

TR 1736

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11NO7
CAS Number:
Molecular Weight:
341.27
EC Number:
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

product name

Aristolochic acid I, powder

Assay

≥90% (HPLC)

form

powder

color

yellow

mp

269-270 °C

solubility

DMSO: soluble
ethanol: soluble

storage temp.

2-8°C

SMILES string

COc1cccc2c1cc([N+]([O-])=O)c3c(cc4OCOc4c23)C(O)=O

InChI

1S/C17H11NO7/c1-23-12-4-2-3-8-9(12)5-11(18(21)22)14-10(17(19)20)6-13-16(15(8)14)25-7-24-13/h2-6H,7H2,1H3,(H,19,20)

InChI key

BBFQZRXNYIEMAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aristolochic acid is a naturally occurring plant metabolite found in Aristolochia sp, Bragantia sp. or Asarum sp. plants. It comprises a mixture of nitrophenanthrene carboxylic acids such as aristolochic acid I and II.

Application

Aristolochic acid I have been used:

  • as a standard for the analysis of Aristolochia sprucei crude extract by high-performance liquid chromatography
  • to study its effects on histone deacetylase 3 (HDAC3) aberration and renal fibrosis
  • to induce acute aristolochic acid nephropathy and to study its impact on miRNA and mRNA expression in mice

Biochem/physiol Actions

Aristolochic acid is a potent inhibitor of phospholipase A2 (PLA2), hyaluronidase, and acetylcholinesterase plasma proteases from snake venoms. Aristolochic acid is considered a herbal medicine and shows therapeutic effects against obstetrics, snake bites, gout, and rheumatism. It exhibits anti-inflammatory and anti-malarial properties. In addition, it is also considered a genotoxic mutagen and causes aristolochic acid nephropathy (AAN), characterized by interstitial fibrosis and urothelial cancer.
Potent phospholipase A2 inhibitor, including calcium ionophore-induced phospholipase A2 activity in neutrophils. Kidney tumor initiator in experimental animal model.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1A - Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongheng Bai et al.
Molecular medicine reports, 16(1), 737-745 (2017-06-01)
Sedum sarmentosum Bunge (SSBE) is a perennial plant widely distributed in Asian countries, and its extract is traditionally used for the treatment of certain inflammatory diseases. Our previous studies demonstrated that SSBE has marked renal anti‑fibrotic effects. However, the underlying molecular
Ziqiang Zhu et al.
Molecular medicine reports, 22(4), 3367-3377 (2020-09-19)
In acute aristolochic acid nephropathy (AAN), aristolochic acid (AA) induces renal injury and tubulointerstitial fibrosis. However, the roles of microRNAs (miRNAs/miRs) and mRNAs involved in AAN are not clearly understood. The aim of the present study was to examine AA‑induced
Ching-Chin Yang et al.
Toxicology, 312, 63-73 (2013-08-14)
Studies have found that ingestion of aristolochic acid (AA) causes nephropathy first by inducing renal tubular cell apoptosis acutely. It is currently unknown whether crosstalk between autophagy and apoptosis orchestrates the fate of tubular cells in acute AA nephropathy. We
Guang-Xing Shui et al.
Evidence-based complementary and alternative medicine : eCAM, 2017, 9536458-9536458 (2017-04-20)
Objectives. The effects of the traditional formula Dahuang Fuzi Decoction (DFD) on chronic aristolochic acid nephropathy (AAN) in mice and its underlying mechanisms were studied. Methods. Mice were randomly divided into the following six groups: the control group, the model
Xiao Y Dai et al.
Oncotarget, 7(10), 10841-10856 (2016-02-26)
Aristolochic acid nephropathy (AAN) is a progressive kidney disease caused by some Chinese herbal medicines, but treatment remains ineffective. Macrophage accumulation is an early feature in human and experimental AAN; however, the role of macrophages in chronic AAN is unknown.

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service