Skip to Content
Merck
All Photos(5)

Key Documents

A4882

Sigma-Aldrich

6-Thioguanine

≥98%

Synonym(s):

2-Amino-6-mercaptopurine, 2-Amino-6-purinethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5S
CAS Number:
Molecular Weight:
167.19
Beilstein:
157765
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

powder

mp

≥300 °C (lit.)

SMILES string

NC1=Nc2nc[nH]c2C(=S)N1

InChI

1S/C5H5N5S/c6-5-9-3-2(4(11)10-5)7-1-8-3/h1H,(H4,6,7,8,9,10,11)

InChI key

WYWHKKSPHMUBEB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

6-Thioguanine is a variant of guanine with hydrogen bonding at the N-7 of the purine ring. Its association with cytosine alters the dimension of the base stacking. 6-Thioguanine usage in treating inflammatory bowel disease (IBD) contributes to nodular regenerative hyperplasia (NRH) in the liver.

Application

6-Thioguanine has been used:
  • to induce autophagy and apoptosis in colorectal cancer cell lines HCT116
  • as a selection marker in the mutation and survival assay in chinese hamster lung fibroblasts culture V79
  • as a selection marker in clonogenic Lung metastasis assay of 4T1-luc cells

Biochem/physiol Actions

Antimetabolite used in the treatment of leukemias. Competes with hypoxanthine and guanine for the enzyme hypoxanthine-guanine phosphoribosyltransferase, which catalzyes a critical step in the generation of purine nucleotides.
Ribosylated and phosphorylated by the same pathway as natural purine bases; as the nucleotide, inhibits a variety of cellular processes involved in nucleic acid synthesis. Has a long history as an effective treatment of leukemia.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Interconnections between apoptotic and autophagic pathways during thiopurine-induced toxicity in cancer cells: the role of reactive oxygen species
Chaabane W and Appell ML
Testing, 7(46), 75616-75616 (2016)
6-thioguanine can cause serious liver injury in inflammatory bowel disease patients
Dubinsky MC, et al.
Gastroenterology, 125(2), 298-303 (2003)
Tumor-suppressor inactivation of GDF11 occurs by precursor sequestration in triple-negative breast cancer
Bajikar SS, et al.
Developmental Cell, 43(4), 418-435 (2017)
Jen-Hwey Chiu et al.
Scientific reports, 10(1), 8060-8060 (2020-05-18)
Our previous study demonstrated that upregulation of multiple epidermal growth factor-like domains 11 (MEGF11) gene expression is involved in the mechanism by which recurrence of Triple Negative Breast Cancer (TNBC) occurs. Our aim was to elucidate the role of MEGF11
Crystal and molecular structure of 6-thioguanine
Bugg CE and Thewalt U
Journal of the American Chemical Society, 92(25), 7441-7445 (1970)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service