Skip to Content
Merck
All Photos(2)

Key Documents

A4362

Sigma-Aldrich

Activin A human

Animal-component free, recombinant, expressed in Nicotiana, >97% (SDS-PAGE), suitable for cell culture

Synonym(s):

Erythroid differentiation protein, Inhibin beta A chain; Activin beta-A chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

product name

Activin A active human, Animal-component free, recombinant, expressed in Nicotiana, >97% (SDS-PAGE)

biological source

human

Quality Level

recombinant

expressed in Nicotiana

Assay

>97% (SDS-PAGE)

form

lyophilized

potency

≤5 ng/mL ED50

mol wt

27.4 kDa (disulfide-linked homodimers of two βA chains, each containing 116 amino residues)

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

Endotoxin, tested (LAL test, < 0.04 EU/ ug protein)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... INHBA(3624)

Amino Acid Sequence

HisHisHisHisHisHisGlyLeuGluCysAspGlyLysValAsnIleCysCysLysLysGlnPhePheValSerPheLysAspIleGlyTrpAsnAspTrpIleIleAlaProSerGlyTyrHisAlaAsnTyrCysGluGlyGluCysProSerHisIleAlaGlyThrSerGlySerSerLeuSerPheHisSerThrValIleAsnHisTyrArgMetArgGlyHisSerProPheAlaAsnLeuLysSerCysCysValProThrLysLeuArgProMetSerMetLeuTyrTyrAspAspGlyGlnAsnIleIleLysLysAspIleGlnAsnMetIleValGluGluCysGlyCysSer

General description

Activins are homodimers or heterodimers of the various β subunit isoforms, belonging to the TGFβ family. Mature Activin A has two 116 amino acids residues βA subunits (βA-βA). Activin exhibits a wide range of biological activities, including mesoderm induction, neural cell differentiation, bone remodelling, haematopoiesis, and reproductive physiology. Activins plays a key role in the production and regulation of hormones such as FSH, LH, GnRH and ACTH. Cells known to express Activin A include fibroblasts, endothelial cells, hepatocytes, vascular smooth muscle cells, macrophages, keratinocytes, osteoclasts, bone marrow monocytes, prostatic epithelium, neurons, chondrocytes, osteoblasts, Leydig cells, Sertoli cells, and ovarian granulosa cells.
As with other members of the super-family, Activins interact with two types of cell surface trans-membrane receptors (Types I and II) which have intrinsic serine/threonine kinase activities in their cytoplasmic domains, Activin type 1 receptors, ACVR1, ACVR1B, ACVR1C and Activin type 2 receptors, ACVR2A, ACVR2B. The biological activity of Activin A can be neutralized by inhibins and by the diffusible TGF-B antagonist, Follistatin.

Physical form

Lyophilized from a Tris HCl 0.05 M buffer at pH 7.4

Reconstitution

Lyophilized protein should be reconstituted in water to a concentration of 50 ng /μl. Due to the protein nature, dimmers and multimers may be observed.

Analysis Note

The biological activity of Activin A is measured by its ability to inhibit mouse plasmacytoma cell line (MPC-11) cells proliferation.
ED50: ≤ 5 ng/mL

Other Notes

Extinction Coefficient: E0.1% = 1.27 (A 280 nm)
p.I: 7.27

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Qian et al.
Biomaterials, 35(36), 9581-9590 (2014-09-06)
Well-defined culture conditions are essential for realizing the full potential of human embryonic stem cells (hESCs) in regenerative medicine where large numbers of cells are required. Synthetic polymers such as poly[2-(methacryloyloxy) ethyl dimethyl-(3-sulfopropyl) ammonium hydroxide] (PMEDSAH), offer multiple advantages over
Yasuhide Ohinata et al.
PloS one, 9(9), e107308-e107308 (2014-09-10)
The inner cell mass (ICM) and trophoblast cell lineages duet early embryonic development in mammals. After implantation, the ICM forms the embryo proper as well as some extraembryonic tissues, whereas the trophoectoderm (TE) exclusively forms the fetal portion of the
Yan Li et al.
The Journal of clinical endocrinology and metabolism, 100(11), E1415-E1427 (2015-08-26)
Activin A increases matrix metalloproteinase (MMP) 2 expression and cell invasion in human trophoblasts, but whether the expression of MMP2 is essential for the proinvasive effect of activin A has yet to be determined. Moreover, the identity of the activin
Melissa L Brown et al.
Islets, 6(5-6), e1017226-e1017226 (2015-04-04)
Emerging evidence suggests that activin with its associated receptors, second messengers, and antagonists would be excellent targets for therapeutic drug development in the treatment of diabetes. We undertook the current study to investigate the ability to extrapolate findings from rodent
Hannah E J Yong et al.
Pregnancy hypertension, 5(4), 346-353 (2015-11-26)
Activin A, a TGFβ family member, circulates in the maternal blood at increasing concentrations throughout gestation during a healthy pregnancy. The circulating concentration of activin A is further increased in pre-eclampsia (PE), a hypertensive disorder of pregnancy that is marked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service