Skip to Content
Merck
All Photos(1)

Key Documents

221M-1

Sigma-Aldrich

Annexin A1 (MRQ-3) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-3, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (221M-14)
vial of 0.5 mL concentrate (221M-15)
bottle of 1.0 mL predilute (221M-17)
vial of 1.0 mL concentrate (221M-16)
bottle of 7.0 mL predilute (221M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

hairy cell leukemia

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

General description

ANXA1 is strongly expressed on the cell membrane and occasionally in the cytoplasm of tumor cells in 97% of samples from patients with hairy cell leukemia. By contrast, B-cell lymphomas other than hairy cell leukemia, including typical splenic lymphoma with villous lymphocytes and patients with variant hairy cell leukemia—as defined by current morphologic, phenotypic, and clinical criteria—are ANXA1-negative. In a study by Falini et al. ANXA1 immunodetection was 100% sensitive and specific for hairy cell leukemia. Normal B cells from different lympho-hemopoietic tissues were ANXA1-negative. In this study the expression of ANXA1 in myeloid cells, macrophages, or T-cell subset served as positive control. These findings validated the results of gene expression profiling in hairy cell leukemia at the protein level by showing that ANXA1 is consistently expressed in this type of leukemic disease, but not in other B-cell lymphomas. Of note is that negativity for ANXA1 was also present in patients with splenic lymphoma with villous lymphocytes, variant hairy cell leukemia, prolymphocytic leukemia, marginal zone and lymphoplasmacytoid lymphomas. Thus, ANXA1 is a molecule specific to hairy cell leukemia that can be used to differentiate this disease from other B-cell lymphomas. Wang et al. showed that high ANXA1 expression is frequent in esophageal and esophagogastric junction adenocarcinomas, is associated with more advanced pathologic T stage and the presence of distant metastasis, and is an independent prognostic factor for patient survival.
Associated products: TRAcP, T-bet.

Quality


IVD

IVD

IVD

RUO

Linkage

Annexin A1 Positive Control Slides, Product No. 221S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu-Hua Xia et al.
Oncogene, 21(43), 6641-6648 (2002-09-21)
The development and progression of human cancer are believed to be due to the alterations of multiple genes or/and their protein products. For identifying the proteins associated with esophageal cancer, we analysed the protein profiles of 24 pairs of esophageal
Kinship structure and health-care improvement in sub-Saharan Africa.
Moses N Katabarwa et al.
Lancet (London, England), 363(9427), 2194-2194 (2004-06-29)
R Dreier et al.
Histochemistry and cell biology, 110(2), 137-148 (1998-08-28)
Annexins constitute a family of Ca2+- and phospholipid-binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. To elucidate a possible correlation of
Kim L Wang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(15), 4598-4604 (2006-08-11)
Annexin A1 (ANXA1) is a calcium-binding protein involved in arachidonic acid metabolism and epidermal growth factor receptor tyrosine kinase pathway. ANXA1 has been implicated in early squamous cell carcinogenesis of esophagus and correlates with degree of tumor differentiation. However, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service