Skip to Content
Merck
All Photos(4)

Key Documents

P3011

Sigma-Aldrich

Potassium thiocyanate

ReagentPlus®, ≥99.0%

Synonym(s):

Potassium rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KSCN
CAS Number:
Molecular Weight:
97.18
Beilstein:
3594799
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥99.0%
form:
powder or crystals

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

powder or crystals

pH

5.3-8.7 (25 °C, 97.2 g/L)

mp

173 °C (lit.)

SMILES string

[K]SC#N

InChI

1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

ZNNZYHKDIALBAK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium thiocyanate (KSCN) is a potassium salt of thiocyanate anion. Its crystalline structure has been analyzed based on polarized infrared spectral data. The inhibition efficiency of KSCN dissolved in 1M HCl against the corrosion of carbon-steel has been evaluated. It facilitates the ring opening of aziridines with high regioselectivity.

Application

Potassium thiocyanate may be used in the synthesis of the following:
  • α-Amino nitriles from tertiary amines.
  • Polysubstituted 2-aminothiazoles from vinyl azides.
  • Benzisothiazol-3(2H)-one derivatives from o-bromobenzamides.
  • Diaryl thioethers from aryl halides.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective ring-opening of aziridines with potassium thiocyanate in the presence of β-cyclodextrin in water.
Reddy MS, et al.
Tetrahedron Letters, 46(38), 6437-6439 (2005)
Synthesis of diaryl thioethers from aryl halides and potassium thiocyanate.
Hajipour AR, et al.
Applied Organometallic Chemistry, 28(12), 879-879 (2014)
Potassium Thiocyanate as Source of Cyanide for the Oxidative α-Cyanation of Tertiary Amines.
Wagner A and Ofial AR.
The Journal of Organic Chemistry, 80(5), 2848-2854 (2015)
Iron (II)-Promoted Synthesis of 2-Aminothiazoles via C=N Bond Formation from Vinyl Azides and Potassium Thiocyanate.
Zhang G, et al.
Advanced Synthesis & Catalysis, 357(5), 1065-1069 (2015)
Polarized infrared spectrum of potassium thiocyanate.
Jones LH.
J. Chem. Phys. , 28(6), 1234-1236 (1958)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service