Skip to Content
Merck
All Photos(1)

Key Documents

676810

Sigma-Aldrich

Ethyl acetate

ACS reagent, ≥99.5%

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

impurities

H2SO4, passes test
≤0.0009 meq/g Titr. acid
≤0.2% water

evapn. residue

≤0.003%

color

APHA: ≤10

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

solubility

alcohol: soluble(lit.)
chloroform: soluble(lit.)

density

0.902 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamics and control of an ethyl acetate reactive distillation column.
Vora N and Daoutidis P.
Industrial & Engineering Chemistry Research, 40(3), 833-849 (2001)
Christer Jansson et al.
Journal of chromatography. A, 1023(1), 93-104 (2004-02-06)
A new multi-residue method for determination of pesticide residues in a wide variety of fruit and vegetables, using the National Food Administration (NFA) ethyl acetate extraction and determination by means of LC-MS/MS, is presented. The method includes pesticides normally detected
Anatase thin films on glass from the chemical vapor deposition of titanium (IV) chloride and ethyl acetate.
O'Neill SA, et al.
Chemistry of Materials, 15(1), 46-50 (2003)
Perspectives for the biotechnological production of ethyl acetate by yeasts.
Loser C, et al.
Applied Microbiology and Biotechnology, 98(12), 5397-5415 (2014)
Manganese oxide OMS-2 as an effective catalyst for total oxidation of ethyl acetate.
Gandhe AR, et al.
Applied Catalysis. B, Environmental, 72(1), 129-135 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service