Skip to Content
Merck
All Photos(1)

Key Documents

650455

Sigma-Aldrich

Cyclohexane

suitable for HPLC, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.06
Assay:
≥99.9%
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

vapor density

2.9 (vs air)

Quality Level

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

500 °F

expl. lim.

9 %

technique(s)

HPLC: suitable

impurities

≤1.0 ppb Fluorescence (quinine) at 365 nm
<0.01% water

evapn. residue

<0.0001%

halogenated residue

<10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 200 nm Amax: 1.00
λ: 225 nm Amax: 0.17
λ: 250 nm Amax: 0.02
λ: 300-400 nm Amax: 0.005

application(s)

food and beverages

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexane, a cycloalkane, is a combustible liquid with petrol like odor. It can be synthesized by hydrogenation of benzene in the presence of nickel or platinum catalysts. Cyclohexane is the precursor for the synthesis of polyamides. Cyclohexane ring forms the structural unit of many naturally occurring products. The conformational changes of cyclohexane have been investigated over the temperature range of 100-1200°C. Conversion of cyclohexane to cyclohexanol and cyclohexanone, via oxidation in the presence of Cu-nanoclusters anchored on nanocrystalline Cr2O3 has been reported.

Application

Suitable for HPLC, spectrophotometry, environmental testing

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 296-296 (1994)
Conformational transitions in cyclohexane and benzol.
Melker AI, et al.
SPIE Proc., 5127 (2003)
Room temperature selective oxidation of cyclohexane over Cu-nanoclusters supported on nanocrystalline Cr2O3.
Sarkar B, et al.
Green Chemistry, 14(9), 2600-2606 (2012)
Amanda L Pitts et al.
Journal of the American Chemical Society, 136(24), 8614-8625 (2014-05-16)
Carbon-hydrogen bond activation reactions of four cycloalkanes (C5H10, C6H12, C7H14, and C8H16) by the Cp'Rh(CO) fragments (Cp' = η(5)-C5H5 (Cp) or η(5)-C5Me5 (Cp*)) were modeled theoretically by combining density functional and coupled cluster theories, and their reaction rates were measured
Sandra Yucra et al.
Environmental health : a global access science source, 7, 59-59 (2008-11-19)
Organophosphates are broad class of chemicals widely used as pesticides throughout the world. We performed a cross-sectional study of associations between dialkylphosphate metabolites of organophosphates and semen quality among pesticide applicators in Majes (Arequipa), Peru. Thirty-one men exposed to organophosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service