Skip to Content
Merck

398608

Sigma-Aldrich

Polyphosphoric acid

~105% H3PO4 basis

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
₩155,803
2.5 L
₩329,672
6 X 500 ML
₩565,152
6 X 2.5 L
₩845,901

₩155,803


Estimated to ship onJanuary 31, 2025Details


To order products, please contact your local dealer.

Select a Size

Change View
500 ML
₩155,803
2.5 L
₩329,672
6 X 500 ML
₩565,152
6 X 2.5 L
₩845,901

About This Item

Linear Formula:
Hn+2PnO3n+1
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38070203
NACRES:
NA.21

₩155,803


Estimated to ship onJanuary 31, 2025Details


To order products, please contact your local dealer.

Assay

~105% H3PO4 basis

Quality Level

form

viscous liquid

bp

300 °C (lit.)

mp

16 °C

density

1.9 g/mL at 25 °C

SMILES string

[P](=O)(O)(O)O.[p]21[o][p]([o][o]2)[o][o]1

InChI

1S/O5P2.H3O4P/c1-2-7-4-3-6(1)5-7;1-5(2,3)4/h;(H3,1,2,3,4)

InChI key

FVYIVXLIMHNYTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
8.075021.07054110132
Pentanal for synthesis

8.08504

Pentanal

solubility

11.7 g/L

solubility

55 g/L

solubility

0.4 g/L

solubility

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

liquid

form

liquid

form

liquid

form

-

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

mp

-91 °C

mp

-8 °C

mp

−130 °C (lit.)

mp

−92 °C (lit.)

General description

Polyphosphoric acid is a hygroscopic, clear and viscous liquid. It has been synthesized by reacting phosphoric acid with phosphorus (V) oxide. It is a moderately strong mineral acid with a wide range of applications.[1] PPA may be used in bone regeneration by treating titanium (Ti) implants.[2]

Application

Polyphosphoric acid may be used in the following processes:
  • Functionalize mesoporous silica–polymer nanocomposites (SBA/PS).[3]
  • As a catalyst in the synthesis of 3,4-dihydropyrimidin-2-(1H)-ones.[4]
  • As a dehydrating agent and phosphorus source in the synthesis of Si, P co-doped carbon (SiPDC) materials.[5]
  • As a catalyst and absorbent of ammonia during the synthesis of dimethyl carbonate (DMC).[6]
  • As a solvent in the synthesis of hyperbranched polybenzoxazoles (HBPBOs).[7]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyphosphoric Acid
Dodd JH.<BR/>
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2001)
Synthesis of hyperbranched polybenzoxazoles and their molecular composites with epoxy resins.
Wu J, et al.
Journal of Applied Polymer Science, 132(18) (2015)
Polyphosphoric acid catalyst for the one-pot synthesis of 3, 4-dihydropyrimidin-2 (1H)-ones by grinding under solvent-free conditions.
Zhao Y, et al.
Indian J. Chem. B, 54(1), 139-141 (2015)
Mesoporous silica polymer nanocomposites encapsulated with polyphosphoric acid for the synthesis of ?-amino carbonyl compounds under environmental benign conditions.
Sachdev D, et al.
New. J. Chem., 39(4), 2633-2641 (2015)
Novel tannin-based Si, P co-doped carbon for supercapacitor applications.
Ramasahayam SK, et al.
Journal of Power Sources, 275, 835-844 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service