Skip to Content
Merck
All Photos(4)

Key Documents

380970

Sigma-Aldrich

Sodium cyanide

reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCN
CAS Number:
Molecular Weight:
49.01
Beilstein:
3587243
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
97%
grade:
reagent grade
form:
powder, chunks or granules
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor density

1.7 (vs air)

vapor pressure

1 mmHg ( 817 °C)

Assay

97%

form

powder, chunks or granules

pH

11-12 (25 °C, 49 g/L)

mp

563.7 °C (lit.)

SMILES string

[Na]C#N

InChI

1S/CN.Na/c1-2;

InChI key

RTVFYQXEHKQMKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium cyanide is a hygroscopic salt and moderately strong base. It is soluble in water. Sodium cyanide is a strong nucleophile used to produce nitriles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen W Kreuzer et al.
Journal of forensic sciences, 57(1), 75-79 (2011-11-02)
Sodium and potassium cyanide are highly toxic, produced in large amounts by the chemical industry, and linked to numerous high-profile crimes. The U.S. Centers for Disease Control and Prevention has identified cyanide as one of the most probable agents to
Asymmetric cyanohydrin formation from aldehydes catalyzed by manganese Schiff base complexes.
Qu Y, et al.
Tetrahedron Asymmetry, 21(2), 187-190 (2010)
Victor M Victor et al.
The Journal of clinical endocrinology and metabolism, 96(10), 3115-3122 (2011-07-23)
Insulin resistance is a feature of polycystic ovary syndrome (PCOS) and is related to mitochondrial and endothelial function. We tested whether hyperandrogenic insulin-resistant women with PCOS, who have an increased risk of vascular disease, display impaired leukocyte-endothelium interactions, and mitochondrial
Heterogeneously Catalyzed Oxidative Cyanation of Tertiary Amines with Sodium Cyanide/Hydrogen Peroxide using Polymer-Supported Iron (II) Phthalocyanines as Catalyst.
Singhal S, et al.
Advanced Synthesis & Catalysis, 352(8), 1338-1344 (2010)
Copper (I) Bromide-Catalyzed Carbonylative Coupling of Aryl Halides with Phenols, Alcohols and Amines using Sodium Cyanide as C1 Source: A Synthesis of Carboxylic Acid Derivatives.
Prasad PK and Sudalai A.
Advanced Synthesis & Catalysis, 356(10), 2231-2238 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service