Skip to Content
Merck
All Photos(1)

Key Documents

Y0001182

Captopril for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Captopril, N-[(S)-3-Mercapto-2-methylpropionyl]-L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO3S
CAS Number:
Molecular Weight:
217.29
Beilstein:
477887
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

captopril

manufacturer/tradename

EDQM

mp

104-108 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H](CS)C(=O)N1CCC[C@H]1C(O)=O

InChI

1S/C9H15NO3S/c1-6(5-14)8(11)10-4-2-3-7(10)9(12)13/h6-7,14H,2-5H2,1H3,(H,12,13)/t6-,7+/m1/s1

InChI key

FAKRSMQSSFJEIM-RQJHMYQMSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Captopril for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Angiotensin converting enzyme inhibitor. Inhibits the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R I Ogilvie et al.
The Canadian journal of cardiology, 14(8), 1025-1033 (1998-09-17)
Twenty-four splenectomized dogs were subjected to rapid right ventricular pacing (RRVP) at 250 beats/min for five weeks. During the final three weeks, four groups six dogs were untreated or treated with captopril alone, with the angiotensin II type 1 (AT1)
Antonio C M Camargo et al.
Toxicon : official journal of the International Society on Toxinology, 59(4), 516-523 (2011-08-13)
The identification of novel endogenous and exogenous molecules acting in the complex mechanism of regulating the vascular tonus has always been of great interest. The discovery of bradykinin (1949) and the bradykinin-potentiating peptides (1965) had a pivotal influence in the
A O Nur et al.
International journal of pharmaceutics, 194(2), 139-146 (2000-02-29)
The development of oral sustained or controlled release dosage form of captopril has been an interested topic of research for a long period of time. Difficulties encountered with such topic based on the fact that the drug is freely water
Javed Mahmood et al.
International journal of radiation oncology, biology, physics, 89(4), 722-728 (2014-05-29)
To investigate the outcome of suppression of the renin angiotensin system using captopril combined with an antioxidant (Eukarion [EUK]-207) for mitigation of radiation-induced lung damage in rats. The thoracic cavity of female Sprague-Dawley rats was irradiated with a single dose
Kyung Hee Chang et al.
Nature communications, 6, 5914-5914 (2015-01-13)
Patients with organ failure of vascular origin have increased circulating haematopoietic stem cells and progenitors (HSC/P). Plasma levels of angiotensin II (Ang-II), are commonly increased in vasculopathies. Hyperangiotensinemia results in activation of a very distinct Ang-II receptor set, Rho family

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service