Skip to Content
Merck
All Photos(1)

Key Documents

Y0001173

Filgrastim

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

filgrastim

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Filgrastim EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ahmet Dirican et al.
Tumori, 99(2), 145-148 (2013-06-12)
Cytotoxic chemotherapy is the basic treatment for metastatic gastric cancer. The "docetaxel, cisplatin, 5-day infusion of 5-FU (DCF5)" regimen is regarded as an effective therapy. However, the poor toxicity profile of this regimen and administration by 5-day infusion are major
I Ia Moiseeva et al.
Voprosy onkologii, 59(2), 71-74 (2013-07-03)
There was studied the influence Dicarbamine and Leykostime on peripheral blood leukocyte composition of rabbits in experimental radiogenic damage to the blood system. Dicarbamine significantly insured the safety of circulating red blood cells, prevented the development of severe postradiation thrombocytopenia
Systemic hypersensitivity reaction mimicking anaphylaxis after first filgrastim administration in a healthy donor.
Ashok Tholpady et al.
Transfusion, 53(5), 1146-1147 (2013-05-11)
Effect of granulocyte colony-stimulating factor on circulating immune and stem cells in septic shock: revisit the basics and consider giving it another chance.
Tomasz Skirecki et al.
American journal of respiratory and critical care medicine, 187(2), 217-219 (2013-01-17)
Kirsi-Maija Kaukonen et al.
BMC infectious diseases, 13, 51-51 (2013-02-01)
Heparin Binding Protein (HBP) is released to blood circulation from activated neutrophils in bacterial infections. It is a potential inducer of vascular leakage and precludes the development of septic shock. Filgrastim induces the production of new neutrophils and modulates their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service