Skip to Content
Merck
All Photos(1)

Key Documents

S7257

Supelco

Sulfamethoxypyridazine

analytical standard

Synonym(s):

N1-(6-Methoxypyridazin-3-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N4O3S
CAS Number:
Molecular Weight:
280.30
Beilstein:
277076
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(NS(=O)(=O)c2ccc(N)cc2)nn1

InChI

1S/C11H12N4O3S/c1-18-11-7-6-10(13-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7H,12H2,1H3,(H,13,15)

InChI key

VLYWMPOKSSWJAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

T P Chorzelski et al.
The American journal of the medical sciences, 306(3), 145-150 (1993-09-01)
This article describes a case of unusual annular erythema-like dermatosis, with histological features of pemphigus foliaceus (subcorneal acantholysis) and IgG antibodies in circulation and bound in vivo to the keratinocyte surface. The reactivity of the antibodies, restricted to human squamous
Ivan Yu Sakharov et al.
Journal of agricultural and food chemistry, 58(6), 3284-3289 (2010-02-24)
An indirect competitive chemiluminescent enzyme-linked immunosorbent assay (CL-ELISA) of sulfamethoxypyridazine (SMP) was developed. The conjugates of streptavidin with cationic horseradish peroxidase (HRP) and anionic soybean peroxidase (SbP) were used in CL-ELISA for the detection of biotinylated anti-SMP antibodies. For streptavidin-HRP
Leticia López-Martínez et al.
Journal of pharmaceutical and biomedical analysis, 30(1), 77-85 (2002-08-02)
The bivariate calibration algorithm was applied to the spectrophotometric simultaneous determination of trimethoprim (TMP), sulfamethoxazole (SMX) or sulphamethoxypyridazine (SMP) binary mixtures in pharmaceutical and veterinary products. The results obtained were compared with those from derivative spectrophotometry. The statistical evaluation of
Sai Ye et al.
Journal of separation science, 30(15), 2360-2369 (2007-09-01)
This paper describes a rapid method for the determination of 14 kinds of sulfonamides (SAs) in wastewater using SPE, and LC-MS/MS with positive ESI (ESI(+)) and selected reaction monitoring (SRM) mode. The SPE was performed on an Oasis hydrophilic-lipophilic-balanced (HLB)
J B Escalera et al.
The Journal of pharmacy and pharmacology, 46(3), 172-176 (1994-03-01)
An approach to reproduce the solubility profile of a drug in several solvent mixtures showing two solubility maxima is proposed in this work. The solubility of sulphamethoxypyridazine was determined at 25 degrees C in several mixtures of varying polarity (hexane:ethyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service