Skip to Content
Merck
All Photos(1)

Key Documents

N7519

Millipore

Nutrient broth

Microbiologically tested, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

Quality Level

sterility

non-sterile

form

powder

composition

Beef extract, 3.0 g/L
Peptone, 5.00 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

application(s)

bioburden testing
environmental
food and beverages
life science and biopharma
water monitoring

suitability

bacteria

Application

Nutrient broth has been used:
  • for culturing Azospirillum brasilense
  • for reconstitution of freeze dried Staphylococcus aureus culture
  • for culturing Pseudomonas aeruginosa NCIMB 8626

Formula variant

With beef extract and peptone.

Preparation Note

Solve 8 g in 1 litre distilled water. Autoclave 15 minutes at 121°C.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Incorporation and controlled release of a hydrophilic antibiotic using poly (lactide-co-glycolide)-based electrospun nanofibrous scaffolds
Kim K, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 98(1), 47-56 (2004)
Michele E Barbour et al.
International journal of nanomedicine, 8, 3507-3519 (2013-10-05)
Chlorhexidine (CHX) is an antimicrobial agent that is efficacious against gram-negative and -positive bacteria and yeasts. Its mechanism of action is based on cell membrane disruption and, as such, it does not promote the development of bacterial resistance, which is
Saikat Jana et al.
NPJ biofilms and microbiomes, 6(1), 19-19 (2020-04-15)
Bacterial biofilms in natural and artificial environments perform a wide array of beneficial or detrimental functions and exhibit resistance to physical as well as chemical perturbations. In dynamic environments, where periodic or aperiodic flows over surfaces are involved, biofilms can
Activity of acetyl-CoA carboxylase is not directly linked to accumulation of lipids when Chlorella vulgaris is co-immobilised with Azospirillum brasilense in alginate under autotrophic and heterotrophic conditions
Leyva LA, et al.
Annals of Microbiology, 65(1), 339-349 (2015)
Oskar A Palacios et al.
Microbial ecology, 77(4), 980-992 (2018-11-07)
The effect of three different nutritional conditions during the initial 12 h of interaction between the microalgae Chlorella sorokiniana UTEX 2714 and the plant growth-promoting bacterium Azospirillum brasilense Cd on formation of synthetic mutualism was assessed by changes in population growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service