Skip to Content
Merck
All Photos(1)

Key Documents

M4141

Sigma-Aldrich

Methanesulfonic acid solution

4 M (with 0.2% (w/v) tryptamine)

Synonym(s):

Sulfomethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3H
CAS Number:
Molecular Weight:
96.11
Beilstein:
1446024
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

concentration

4 M (with 0.2% (w/v) tryptamine)

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O

InChI

1S/CH4O3S/c1-5(2,3)4/h1H3,(H,2,3,4)

InChI key

AFVFQIVMOAPDHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For complete protein and peptide hydrolysis with tryptophan recovery. After hydrolysis the samples are diluted prior to amino acid analysis.

Packaging

Ampules sealed under argon.

Caution

air sensitive, store under inert conditions

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Gulfam Ahmad et al.
Frontiers in pharmacology, 11, 556020-556020 (2020-10-13)
Chronic inflammatory bowel disease (IBD) is a condition with multifactorial pathophysiology. To date, there is no permanent cure and the disease is primarily managed by immunosuppressive drugs; long-term use promotes serious side effects including increased risk malignancies. The current study
Luke F Gamon et al.
Redox biology, 36, 101586-101586 (2020-06-07)
The precise characterization and quantification of oxidative protein damage is a significant challenge due to the low abundance, large variety, and heterogeneity of modifications. Mass spectrometry (MS)-based techniques at the peptide level (proteomics) provide a detailed but limited picture due
Renata Solarska et al.
Nanoscale, 4(5), 1553-1556 (2012-02-01)
Nanostructuring of semiconductor films offers the potential means for producing photoelectrodes with improved minority charge carrier collection. Crucial to the effective operation of the photoelectrode is also the choice of a suitable electrolyte. The behaviour of the nanostructured WO(3) photoanodes
Solvejg Jørgensen et al.
Physical chemistry chemical physics : PCCP, 15(14), 5140-5150 (2013-03-02)
The gas phase reaction between methane sulfonic acid (CH3SO3H; MSA) and the hydroxyl radical (HO), without and with a water molecule, was investigated with DFT-B3LYP and CCSD(T)-F12 methods. For the bare reaction we have found two reaction mechanisms, involving proton
Chris Twelves et al.
Breast cancer research and treatment, 148(3), 553-561 (2014-11-09)
Data from two phase 3 studies of eribulin were pooled in analyses initially requested by the European Medicines Agency to assess whether specific patient subgroups, previously treated with an anthracycline and a taxane, benefited from eribulin. Study 305/EMBRACE included women

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service