Skip to Content
Merck
All Photos(1)

Documents

M1880000

Miconazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14Cl4N2O
CAS Number:
Molecular Weight:
416.13
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

miconazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C18H14Cl4N2O/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22/h1-8,11,18H,9-10H2

InChI key

BYBLEWFAAKGYCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Miconazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Stevens et al.
The American journal of medicine, 60(2), 191-202 (1976-02-01)
Fourteen patients with chronic coccidioidomycosis, many of whom had complicating concurrent diseases and/or had failed to respond to amphotericin therapy, were treated with intravenous miconazole, a synthetic imidazole drug previously shown to be effective in experimental murine coccidioidomycosis. Up to
R C Heel et al.
Drugs, 19(1), 7-30 (1980-01-01)
Miconazole is an imidazole antifungal drug which has recently become available for systemic use. Its antifungal activity has been well studied and it is active in vitro against a wide range of fungi. Published and unpublished reports of the use
Pascale Quatresooz et al.
Expert opinion on pharmacotherapy, 9(11), 1927-1934 (2008-07-17)
Miconazole nitrate is a time-honored antifungal of the imidazole class. To revisit the various aspects of action of the drug in a dermatologic setting. Review of the current peer-reviewed publications. Miconazole essentially inhibits 14alpha-demethylase, an enzyme required for the biosynthesis
J Symoens
Proceedings of the Royal Society of Medicine, 70 Suppl 1, 4-8 (1977-01-01)
Intravenous treatment with miconazole brought about the recovery of 90% of patients with gastrointestinal or systemic candidosis. Miconazole given by the same route has also been found effective in the treatment of cryptococcosis, coccidioidomycosis, and paracoccidioidomycosis. Cryptococcal and coccidioidal meningitis
D A Stevens
Drugs, 26(4), 347-354 (1983-10-01)
Intravenous miconazole can produce responses in patients with various manifestations of coccidioidal disease, even if they have failed to respond to amphotericin B. In 4 large series of 33, 33, 46 and 31 courses of miconazole for skin and soft

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service