Skip to Content
Merck
All Photos(3)

Documents

G5767

Sigma-Aldrich

D-(+)-Glucose

ACS reagent

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

wheat

Quality Level

grade

ACS reagent

form

powder

optical activity

[α]25/D +52.5 to +53.0°(lit.)

impurities

Starch, passes test
≤0.002 meq/g Titratable acid
≤0.005% Insoluble matter

ign. residue

≤0.02%

loss

≤0.2% loss on drying, 105°C

color

colorless

mp

150-152 °C (lit.)

anion traces

chloride (Cl-): ≤0.01%
sulfate, sulfite (as SO42-): ≤0.005%

cation traces

Fe: ≤5 ppm
heavy metals (as Pb): ≤5 ppm

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

General description

D-glucose, an aldohexose, possesses four chiral carbon atoms. It is an important product obtained from starch hydrolyzed by acid and/or enzymes.

Application

D-(+)-Glucose has been used:
  • as a supplement in Dulbecco′s phosphate-buffered saline (DPBS) for standardized serum bactericidal assay (SBA) using human complement
  • as a component in 4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid (HEPES)-buffered Tyrode′s solution to treat the excised diaphragmatic specimens to assess the effect of osmolarity changes on the intrinsic lymphatic pumping mechanism
  • to study the effect of restoring circulating glucose content in insulin-responsive neurons of rats
  • for oral glucose tolerance tests (OGTTs)

Biochem/physiol Actions

Glucose, the universal energy source for all human cells, is the vital precursor for the production and conversion of several downstream carbohydrates. The uptake of glucose is a key indicator of cellular energy metabolism. Glucose infusion can block the release of oxytocin and vasopressin by insulin-induced hypoglycemia in humans.

Preparation Note

The material may produce a yellowish solution if prepared at a higher concentration than 2 g + 15mL.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jashanpreet Kaur et al.
PloS one, 10(3), e0122354-e0122354 (2015-03-27)
Genotyping studies of Australian Scedosporium isolates have revealed the strong prevalence of a recently described species: Scedosporium aurantiacum. In addition to occurring in the environment, this fungus is also known to colonise the respiratory tracts of cystic fibrosis (CF) patients.
Aïcha Gharbi et al.
AMB Express, 5, 9-9 (2015-04-09)
Carvacrol, an aromatic terpenic compound, known to be antimicrobial was grafted onto gold surfaces via two strategies based on newly-synthesized cross-linkers involving either an ester bond which can be cleaved by microbial esterases, or a covalent ether link. Surface functionalizations
Yu-ting Alex Chiang et al.
American journal of physiology. Endocrinology and metabolism, 306(7), E707-E722 (2013-12-26)
p21-Activated protein kinases (PAKs) are centrally involved in a plethora of cellular processes and functions. Their function as effectors of small GTPases Rac1 and Cdc42 has been extensively studied during the past two decades, particularly in the realms of cell
Lars P Bechmann et al.
Journal of hepatology, 56(4), 952-964 (2011-12-17)
It is widely known that the liver is a central organ in lipogenesis, gluconeogenesis and cholesterol metabolism. However, over the last decades, a variety of pathological conditions highlighted the importance of metabolic functions within the diseased liver. As observed in
Dov B Ballak et al.
Nature communications, 5, 4711-4711 (2014-09-04)
Cytokines of the IL-1 family are important modulators of obesity-induced inflammation and the development of systemic insulin resistance. Here we show that IL-1 family member IL-37, recently characterized as an anti-inflammatory cytokine, ameliorates obesity-induced inflammation and insulin resistance. Mice transgenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service