Skip to Content
Merck
All Photos(1)

Documents

E0050000

Econazole nitrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-[2-[(4-Chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole mononitrate, 1-{2,4-Dichloro-β-[(p-chlorobenzyl)oxy}phenethyl]imidazole mononitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16Cl3N3O4
CAS Number:
Molecular Weight:
444.70
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

econazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C18H15Cl3N2O.HNO3/c19-14-3-1-13(2-4-14)11-24-18(10-23-8-7-22-12-23)16-6-5-15(20)9-17(16)21;2-1(3)4/h1-9,12,18H,10-11H2;(H,2,3,4)

InChI key

DDXORDQKGIZAME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Econazole nitrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Peng Yan et al.
Archives of pharmacal research, 34(6), 1007-1013 (2011-07-05)
Osteoclasts are primary bone resorption cells and intervention in osteoclast activation is considered an effective therapeutic approach to treatment of bone diseases involving osteoclasts. TRPV5 was detected in osteoclasts and it has been thought to take part in the transportation
Xianzhi Peng et al.
The Science of the total environment, 426, 311-317 (2012-04-24)
Residue of azole antifungals in the environment is of concern due to the environmental risks and persistence. Distribution, behavior, and fate of frequently used azole antifungal pharmaceuticals were investigated in wastewater at two sewage treatment plants (STPs) in China. Fluconazole
Poonam Verma et al.
Nanomedicine : nanotechnology, biology, and medicine, 8(4), 489-496 (2011-08-16)
This project aimed at developing nanovesicles of econazole nitrate (EN) and formulating them as a suitable dermatological gel for improved therapeutic efficacy, better dispersity, and good storage stability. Ethosomes were prepared by cold method and evaluated for the mean diameter
Azza A Mahmoud et al.
International journal of pharmaceutics, 413(1-2), 229-236 (2011-05-05)
Development of efficient ocular delivery nanosystems remains a major challenge to achieve sustained therapeutic effect. The purpose of this work was to develop chitosan nanoparticles using sulfobutylether-β-cyclodextrin (SBE-β-CD) as polyanionic crosslinker and to investigate the potential of using those nanostructures
Periocular rash.
Huan J Chang
JAMA, 306(11), 1263-1264 (2011-09-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service