Skip to Content
Merck
All Photos(1)

Key Documents

CCW4111

Millipore

SGQ Broth in tubes

Liquid medium in Tubes for fungi and yeasts, especially for Dekkera spp., suitable for microbiology

Synonym(s):

SGQB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Quality Level

form

suspension

shelf life

12 mo. at 8‑25

feature

ready-to-use

packaging

box of 20 tubes

storage condition

(Tightly closed)

parameter

10 mL filling volume

color

slightly yellowish

pH

4.3±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

lactic acid bacteria

General description

The SGQ Broth allows the growth of organisms that can spoil low-acid beverages, specially Dekkera spp.

Application

SGQ Broth in tubes is used to detect low-acid beverage-spoiling microorganisms, specially Dekkera spp.

Components

  • 1 box with 20 tubes
  • 16x113 mm glass tubes, ink labeled

Other Notes

  • Liquid medium formulated according to the specifications of the customer.
  • Use the medium according to the intended purpose, samples, and validated methods.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service