Skip to Content
Merck
All Photos(3)

Key Documents

C9481

Sigma-Aldrich

Carboxymethylcellulose sodium

meets USP testing specifications, Medium viscosity

Synonym(s):

Carboxymethylcellulose sodium salt, Sodium carboxymethylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

6.5-9.5% anhydrous basis (Sodium)

form

powder

color

light yellow

pH

(10 g/L68 °F neutral)

viscosity

400-800 cP, 2 % in H2O(25 °C)(lit.)

mp

> 300.05  °C ((> 572.09 °F ))

solubility

water: soluble

SMILES string

[Na].OC(C(O)C(O)C=O)C(O)CO.OC(=O)C

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carboxymethylcellulose sodium is a viscous polysaccharide that belongs to a high molecular weight category. It has mucoadhesive property and is used during eye surgery. Carboxymethylcellulose sodium promotes re-epithelialization of the epithelial cells in corneal wounds. It is useful for the study of attached cell and three-dimensional tissue culture models.

Application

Carboxymethylcellulose sodium has been used as a vehicle for tamoxifen citrate, sorafenib, and savolitinib. It has also been used as a component of serum-free medium (SFM) for the suspension of human umbilical vein endothelial spheroids.
Useful for the study of attached cell and three-dimensional tissue culture models.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances in modified cellulose for tissue culture applications
Courtenay J, et al.
Molecules (Basel), 23(3), 654-654 (2018)
Carboxymethylcellulose binds to human corneal epithelial cells and is a modulator of corneal epithelial wound healing
Garrett Q, et al.
Investigative Ophthalmology & Visual Science, 48(4), 1559-1567 (2007)
Anti-estrogen resistance in human breast tumors is driven by JAG1-NOTCH4-dependent cancer stem cell activity
Simoes BM, et al.
Testing, 12(12), 1968-1977 (2015)
Vascular CXCR4 expression promotes vessel sprouting and sensitivity to sorafenib treatment in hepatocellular carcinoma
Xu J, et al.
Clinical Cancer Research, 23(15), 4482-4492 (2017)
The MET inhibitor AZD6094 (Savolitinib, HMPL-504) induces regression in papillary renal cell carcinoma patient-derived xenograft models
Schuller AG, et al.
Clinical Cancer Research, 21(12), 2811-2819 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service