Skip to Content
Merck
All Photos(1)

Key Documents

B0500000

Benzathine benzylpenicillin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36N4O8S2 · C16H20N2
CAS Number:
Molecular Weight:
909.12
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

penicillin & derivatives

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/2C16H18N2O4S.C16H20N2/c2*1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-3-7-15(8-4-1)13-17-11-12-18-14-16-9-5-2-6-10-16/h2*3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);1-10,17-18H,11-14H2/t2*11-,12+,14-;/m11./s1

InChI key

BVGLIYRKPOITBQ-ANPZCEIESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzathine benzylpenicillin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

WHO Model Prescribing Information: Drugs Used in Bacterial Infections
WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue (2001)
Clavi syphilitici--an unusual presentation of syphilis.
Catarina Moreira et al.
Journal of the American Academy of Dermatology, 70(6), e131-e132 (2014-05-17)
Ahmadreza Moradi et al.
American journal of ophthalmology, 159(2), 334-343 (2014-12-03)
To describe the clinical outcomes of ocular syphilis. Retrospective chart review. The charts of patients with ocular syphilis (regardless of human immunodeficiency virus [HIV] status) seen in a uveitis referral center between 1984 and 2014 were reviewed. The study included
Treatment of late-stage syphilis.
Aram S A van Brussel et al.
JAMA, 313(9), 968-969 (2015-03-04)
Treatment of late-stage syphilis--reply.
Meredith E Clement et al.
JAMA, 313(9), 969-970 (2015-03-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service