Skip to Content
Merck
All Photos(1)

Key Documents

95621

Sigma-Aldrich

Chelex® 100 sodium form

200-400 mesh particle size

Synonym(s):

crosslinked polystyrene-iminodiacetate ion exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.22

product line

Chelex®

Quality Level

form

powder or chunks

reaction suitability

reaction type: solution phase peptide synthesis

loss

~80% loss on drying, 110°C

matrix

polystyrene/DVB

matrix active group

paired iminodiacetate groups

particle size

200-400 mesh

Looking for similar products? Visit Product Comparison Guide

General description

Chelex®100 sodium form is a chelating resin made of a styrene and divinylbenzene copolymer. It has a pair of iminodiacetate ions that chelate polyvalent metal ions, shows greater metal ion selectivity, and has a stronger binding force.

Application

Chelex®100 sodium form is commonly used:
  • As an ion exchange resin, in wastewater treatment due to its high affinity for metal ions
  • For the extraction and purification of DNA
  • As adsorbent in protein purification
  • In determination of trace elements in marine water

Legal Information

Chelex is a registered trademark of Bio-Rad Laboratories, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion-exchange of nickel by iminodiacetic acid chelating resin Chelex 100
H Leinonen, et al.
Reactive and Functional Polymers, 43, 1-6 (2000)
Some examples of the use of amphoteric ion-exchange resins for inorganic separations.
Samczynski Z and Dybczynski R.
Journal of Chromatography A, 789(1), 157-167 (1997)
Henrik H De Fine Licht et al.
BMC evolutionary biology, 14, 244-244 (2014-12-05)
Cooperative benefits of mutualistic interactions are affected by genetic variation among the interacting partners, which may have consequences for interaction-specificities across guilds of sympatric species with similar mutualistic life histories. The gardens of fungus-growing (attine) ants produce carbohydrate active enzymes
Standing wave design of carousel ion-exchange processes for the removal of zinc ions from a protein mixture
S Mun, et al.
Industrial & Engineering Chemistry Research, 45, 316-329 (2006)
One-step purification/extraction method to access glyphosate, glufosinate, and their metabolites in natural waters
CT Vu, et al.
Journal of Chromatography A, 1649, 462188-462188 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service