Skip to Content
Merck
All Photos(2)

Documents

91242

Supelco

Sulfluramid

PESTANAL®, analytical standard

Synonym(s):

N-Ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-octanesulfonamide, N-Ethylperfluorooctane-1-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6F17NO2S
CAS Number:
Molecular Weight:
527.20
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 22.3-23.3 wt. %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C10H6F17NO2S/c1-2-28-31(29,30)10(26,27)8(21,22)6(17,18)4(13,14)3(11,12)5(15,16)7(19,20)9(23,24)25/h28H,2H2,1H3

InChI key

CCEKAJIANROZEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfluramid is a slow-acting insecticide for controlling ants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfluramid may be used as a reference standard for the determination of sulfluramid and its isomers in ant bait by capillary column gas chromatography.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R G Schnellmann et al.
Biochimica et biophysica acta, 1016(3), 344-348 (1990-04-26)
The effects of sulfluramide (N-ethylperfluorooctane sulfonamide) and perfluorooctane sulfonamide (DESFA) on isolated rabbit renal cortical mitochondria (RCM) were examined. Sulfluramid (1-100 microM) and DESFA (0.5-50 microM) increased state 4 respiration of RCM respiring on pyruvate/malate or succinate in a concentration
Comparison of baits containing fipronil and sulfluramid for the control of Vespula wasps.
Harris R J, et al.
New Zealand Journal of Zoology, 28(1), 39-48 (2001)
R O Manning et al.
Drug metabolism and disposition: the biological fate of chemicals, 19(1), 205-211 (1991-01-01)
The objectives of this study were to characterize the absorption, distribution, and elimination of sulfluramid (N-ethyl perfluorooctane sulfonamide) and its major metabolite, perfluorooctane sulfonamide (DESFA), in order to assess the effect of dosage vehicle on their pharmacokinetics. In Trial 1
Margie M Peden-Adams et al.
Journal of toxicology and environmental health. Part A, 70(13), 1130-1141 (2007-06-15)
Perfluorinated hydrocarbons have been manufactured for over 40 yr and have numerous applications in industry. This group of compounds has recently generated much interest, as some of these compounds such as perfluorooctane sulfonate (PFOS) and perfluoroctanic acid (PFOA) are persistent
B L Reid et al.
Journal of economic entomology, 83(1), 148-152 (1990-02-01)
The LD50 of sulfluramid topically applied to 2-d-old, fifth instars of the German cockroach, Blattella germanica (L.), was estimated at 14.5 micrograms/g (95% FL = 13.7-15.4 micrograms/g). Sulfluramid was significantly more toxic than topically applied hydramethylnon (LD50 = 29.2 [19.0-46.5]

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service