Skip to Content
Merck
All Photos(2)

Documents

80345

Supelco

Tyramine

analytical standard

Synonym(s):

2-(4-Hydroxyphenyl)ethylamine, 4-(2-Aminoethyl)phenol, 4-Hydroxyphenethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
137.18
Beilstein:
1099914
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)
98.0-102.0% (wt., NT)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

175-181 °C/8 mmHg (lit.)

mp

160-162 °C (lit.)

suitability

conforms to structure for Infrared spectrum

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

NCCc1ccc(O)cc1

InChI

1S/C8H11NO/c9-6-5-7-1-3-8(10)4-2-7/h1-4,10H,5-6,9H2

InChI key

DZGWFCGJZKJUFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Can enter catecholaminergic terminals and be released as a false transmitter.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Roeder et al.
Archives of insect biochemistry and physiology, 54(1), 1-13 (2003-08-28)
The phenolamines tyramine and octopamine are decarboxylation products of the amino acid tyrosine. Although tyramine is the biological precursor of octopamine, both compounds are independent neurotransmitters, acting through various G-protein coupled receptors. Especially, octopamine modulates a plethora of behaviors, peripheral
Thomas Roeder
Annual review of entomology, 50, 447-477 (2004-09-10)
Octopamine (OA) and tyramine (TA) are the invertebrate counterparts of the vertebrate adrenergic transmitters. They are decarboxylation products of the amino acid tyrosine, with TA as the biological precursor of OA. Nevertheless, both compounds are independent neurotransmitters that act through
Moussa B H Youdim et al.
Neurotoxicology, 25(1-2), 243-250 (2003-12-31)
The major side effect with the use of first generation of non selective monoamine oxidase (MAO) inhibitors as neuropsychiatric drugs was what became known as the "cheese reaction". Namely, potentiation of sympathomimetic activity of ingested tyramine present in cheese and
Jack J Chen et al.
Journal of clinical pharmacology, 52(5), 620-628 (2011-06-02)
Rasagiline is an irreversible monoamine oxidase type B (MAO-B) inhibitor indicated for the treatment of the signs and symptoms of idiopathic Parkinson disease as initial monotherapy and as adjunct therapy to levodopa. Pharmacologic inhibition of monoamine oxidase type A (MAO-A)
The tyramine conjugation test as a trait marker for endogenous unipolar depression and a predictor of treatment response.
J Jarman
Molecular aspects of medicine, 13(3), 249-261 (1992-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service