Skip to Content
Merck
All Photos(1)

Key Documents

74777

Millipore

Astragaloside IV

>98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H68O14
CAS Number:
Molecular Weight:
784.97
MDL number:
UNSPSC Code:
41106200
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥98.0% (TLC)
>98.0%

form

powder

mp

295-296 °C (lit.)

SMILES string

CC(C)(O)[C@@H]1CC[C@@](C)(O1)[C@H]2[C@@H](O)C[C@@]3(C)[C@@H]4C[C@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)[C@H]6C(C)(C)[C@H](CC[C@@]67C[C@@]47CC[C@]23C)O[C@@H]8OC[C@@H](O)[C@H](O)[C@H]8O

InChI

1S/C41H68O14/c1-35(2)24(54-33-29(48)26(45)20(44)17-51-33)9-11-41-18-40(41)13-12-37(5)31(39(7)10-8-25(55-39)36(3,4)50)19(43)15-38(37,6)23(40)14-21(32(35)41)52-34-30(49)28(47)27(46)22(16-42)53-34/h19-34,42-50H,8-18H2,1-7H3/t19-,20+,21-,22+,23-,24-,25-,26-,27+,28-,29+,30+,31-,32-,33-,34+,37+,38-,39+,40-,41+/m0/s1

InChI key

QMNWISYXSJWHRY-YLNUDOOFSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anti-inflammatory.

Other Notes

Chemical constituent of Astragali radix

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenya Shan et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 37(1), 85-88 (2012-06-30)
To provide a scientific basis for the drug-combination and aim to examine whether astragaloside IV has the impact on the cytochrome P450 enzymes. Tolbutamide, chlorzoxazone, coumarin, nifedipine, and phenacetin were as probe substrates of rat CYP2C9, CYP2E1, CYP2A6, CYP3A4, and
Dingkun Gui et al.
PloS one, 7(6), e39824-e39824 (2012-06-30)
Glucose-induced reactive oxygen species (ROS) production initiates podocyte apoptosis, which represents a novel early mechanism leading to diabetic nephropathy (DN). Here, we tested the hypothesis that Astragaloside IV(AS-IV) exerts antioxidant and antiapoptotic effects on podocytes under diabetic conditions. Apoptosis, albuminuria
I. Kitagawa et al.
Chemical & Pharmaceutical Bulletin, 31, 698-698 (1983)
Ming Hao et al.
Zhonghua yi xue za zhi, 92(30), 2104-2107 (2012-11-20)
To explore the protective effects of astragaloside IV on retinal ganglion cells (RGC) incubated under high glucose. Cultured RGC-5 were divided into 3 groups: control, high glucose and high glucose+astragaloside IV. The survival rate of cell was measured by CCK-8
Li-Hua Peng et al.
Biological & pharmaceutical bulletin, 35(6), 881-888 (2012-06-13)
Topical delivery of therapeutic agents at the time of injury to accelerate skin repair and prevent the formation of scars during the wound healing process has received increasing attention and represents a novel regenerative and prophylactic strategy for wound treatment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service