Skip to Content
Merck
All Photos(1)

Key Documents

45645

Supelco

Propyzamide

PESTANAL®, analytical standard

Synonym(s):

Pronamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11Cl2NO
CAS Number:
Molecular Weight:
256.13
Beilstein:
882391
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(C)(NC(=O)c1cc(Cl)cc(Cl)c1)C#C

InChI

1S/C12H11Cl2NO/c1-4-12(2,3)15-11(16)8-5-9(13)7-10(14)6-8/h1,5-7H,2-3H3,(H,15,16)

InChI key

PHNUZKMIPFFYSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propyzamide, a well-known herbicide, belongs to the benzamide class of compounds.

Application

Propyzamide may be used as a reference standard for the analysis of propyzamide in:
  • Chlorophyll-containing matrices by dispersive-solid phase extraction (d-SPE) and gas chromatography-tandem mass spectrometry (GC-MS/MS).
  • Fruits and vegetables by GC equipped with Q Extractive Orbitrap consisting of electron ionization (EI) source and hybrid quadrupole Orbitrap mass spectrometer.
  • Surface water by liquid chromatography along with MS/MS with electrospray ionization (ESI) in multiple reaction monitoring (MRM) mode.
  • Honeybees and shellfish} by d-SPE followed by analyses using LC-MS/MS as well as GC-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Complete mineralization of propyzamide in aqueous solution containing TiO2 particles and H2O2 by the simultaneous irradiation of light and ultrasonic waves.
Yano J, et al.
Ultrasonics Sonochemistry, 12(3), 197-203 (2005)
A multi-component method to determine pesticides in surface water by liquid-chromatography tandem quadrupole mass spectrometry.
Ramos AM, et al.
Water and environment journal : the journal, 31(3), 380-387 (2017)
Low concentrations of propyzamide and oryzalin alter microtubule dynamics in Arabidopsis epidermal cells.
Nakamura M, et al.
Plant & Cell Physiol., 45(9), 1330-1334 (2004)
Multi-residue method for the determination of pesticides and pesticide metabolites in honeybees by liquid and gas chromatography coupled with tandem mass spectrometry?Honeybee poisoning incidents.
Kiljanek T, et al.
Journal of Chromatography A, 1435(3), 100-114 (2016)
Shifting the paradigm in gas chromatography mass spectrometry pesticide analysis using high resolution accurate mass spectrometry.
Ucles S, et al.
Journal of Chromatography A, 1501(9), 107-116 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service