Skip to Content
Merck
All Photos(1)

Documents

45478

Supelco

Ethirimol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19N3O
CAS Number:
Molecular Weight:
209.29
Beilstein:
882476
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCc1c(C)nc(NCC)nc1O

InChI

1S/C11H19N3O/c1-4-6-7-9-8(3)13-11(12-5-2)14-10(9)15/h4-7H2,1-3H3,(H2,12,13,14,15)

InChI key

BBXXLROWFHWFQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Proença et al.
Forensic science international, 133(1-2), 95-100 (2003-05-14)
Fenarimol (Rubigan) is a pyrimidine ergosterol biosynthesis inhibitor used as a systemic fungicide. The authors present a fatal fenarimol intoxication case analysed in the Forensic Toxicology Service of the National Institute of Legal Medicine. The results were used to compare
[Chromatographic determination of Milgo in water, soil, and plants].
A A Krasnykh et al.
Gigiena i sanitariia, (6)(6), 69-70 (1980-06-01)
L J Fitzpatrick et al.
Journal of chromatography. A, 918(2), 429-433 (2001-06-16)
This paper assesses the effect of pressurised fluid extraction (PFE) on the recovery of bupirimate and its degradation product, ethirimol from a range of soil types. The analytes were extracted under standard conditions (pressure, 2000 p.s.i.; temperature, 100 degrees C;
Shagufta Khan et al.
Food chemistry, 199, 870-875 (2016-01-19)
In the present communication, a non-covalent fenarimol-imprinted polymer was synthesized by precipitation polymerization technique using methacrylic acid (MAA) as a functional monomer, ethylene glycol dimethacrylate (EGDMA) as a cross-linker, and azobisisobutyronitrile (AIBN) as an initiator in different porogenic solvent. Binding
Alexander I Vardavas et al.
Mutation research, 829-830, 26-32 (2018-05-01)
Imidacloprid (IMI) is a systemic, chloro-nicotinyl insecticide classified in Regulation N° 1272/2008 of the European Commision as "harmful if swallowed and very toxic to aquatic life, with long-lasting effects". IMI is metabolized in vitro both by aldehyde oxidase (AOX) (reduction)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service