Skip to Content
Merck
All Photos(1)

Key Documents

43534

Supelco

Glycitein

analytical standard

Synonym(s):

4′,7-Dihydroxy-6-methoxyisoflavone, Glycetein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc2C(=O)C(=COc2cc1O)c3ccc(O)cc3

InChI

1S/C16H12O5/c1-20-15-6-11-14(7-13(15)18)21-8-12(16(11)19)9-2-4-10(17)5-3-9/h2-8,17-18H,1H3

InChI key

DXYUAIFZCFRPTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycitein is an isoflavone found in soy food products.[1]

Application

Glycitein may be used as a reference standard in the determination of glycitein in hydrolyzed dry soya extracts using reversed-phase high-performance liquid chromatography (RP-HPLC).[2]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a RP-HPLC method for quantification of isoflavone aglycones in hydrolyzed soy dry extracts.
CCdI, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 836(1-2), 74-78 (2006)
Estrogenic activity of glycitein, a soy isoflavone
Song T T, et al.
Journal of Agricultural and Food Chemistry, 47(4), 1607-1610 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service