Skip to Content
Merck
All Photos(2)

Key Documents

43504

Supelco

O-Ethylhydroxylamine hydrochloride

for GC derivatization, LiChropur

Synonym(s):

Ethoxyamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2ONH2 · HCl
CAS Number:
Molecular Weight:
97.54
Beilstein:
3606639
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

99.0-101.0% (wt., AT)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

technique(s)

gas chromatography (GC): suitable

mp

130-133 °C (lit.)

SMILES string

Cl[H].CCON

InChI

1S/C2H7NO.ClH/c1-2-4-3;/h2-3H2,1H3;1H

InChI key

NUXCOKIYARRTDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

O-Ethylhydroxylamine hydrochloride may be used as a derivatization reagent in the following:
  • Separation and analysis of complex monosaccharide mixtures using chromatography techniques.
  • Analysis of carbohydrates in complex matrices of biological and synthetic origin using gas chromatography coupled to mass spectrometry (GC-MS).

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of an advanced derivatization protocol for the unambiguous identification of monosaccharides in complex mixtures by gas and liquid chromatography
Haas M, et al.
Journal of Chromatography A, 1568, 160-167 (2018)
Rossella Grande et al.
Frontiers in microbiology, 11, 1705-1705 (2020-08-28)
Helicobacter pylori colonizes approximately 50% of the world's population, and it is the cause of chronic gastritis, peptic ulcer disease, and gastric cancer. The increase of antibiotic resistance is one of the biggest challenges of our century due to its
A I Ruiz-Matute et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(17-18), 1226-1240 (2010-12-28)
GC and GC-MS are excellent techniques for the analysis of carbohydrates; nevertheless the preparation of adequate derivatives is necessary. The different functional groups that can be found and the diversity of samples require specific methods. This review aims to collect
M Becker et al.
Journal of chromatography. A, 1281, 115-126 (2013-02-13)
Gas chromatographic analysis of complex carbohydrate mixtures requires highly effective and reliable derivatisation strategies for successful separation, identification, and quantitation of all constituents. Different single-step (per-trimethylsilylation, isopropylidenation) and two-step approaches (ethoximation-trimethylsilylation, ethoximation-trifluoroacetylation, benzoximation-trimethylsilylation, benzoximation-trifluoroacetylation) have been comprehensively studied with regard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service