Skip to Content
Merck
All Photos(3)

Key Documents

37520

Sigma-Aldrich

3,4-Dihydroxybenzaldehyde

purum, ≥97.0% (HPLC)

Synonym(s):

Protocatechualdehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CHO
CAS Number:
Molecular Weight:
138.12
Beilstein:
774381
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (HPLC)

form

powder

mp

150-155 °C
150-157 °C (lit.)

SMILES string

Oc1ccc(C=O)cc1O

InChI

1S/C7H6O3/c8-4-5-1-2-6(9)7(10)3-5/h1-4,9-10H

InChI key

IBGBGRVKPALMCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dihydroxybenzaldehyde has been recognized as one of the antifungal compound extracted from the outer skin of green Cavendish bananas. It can be synthesized from catechol via Fries rearrangement.
3,4-Dihydroxybenzaldehyde is reported as bioactive compound which inhibits the H2O2-induced apoptosis of granulosa cells. Oxidation of 3,4-dihydroxybenzaldehyde on glassy carbon electrodes is reported to afford stable redox-active electropolymerized films containing a quinone moity.

Application

3,4-Dihydroxybenzaldehyde (Protocatechualdehyde) may be employed as starting reagent for the synthesis of 4-vinylbenzocrown ether.
3,4-Dihydroxybenzaldehyde may be used for the surface modification of nanocrystalline TiO2 particles. Electrodeposited layer of 3,4-dihydroxybenzaldehyde may be used as effective redox mediator during oxidation of NADH at graphene. It may be used in the preparation of new diSchiff base ligands, which forms di-, tri- and tetranuclear Co(II) and Cu(II) complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahlam Jameel Abdulghani et al.
Bioinorganic chemistry and applications, 2013, 219356-219356 (2014-01-24)
A series of new di-, tri-, and tetranuclear Co(II) and Cu(II) complexes of three new diSchiff base ligands were synthesized by two different methods. The first method involved the synthesis of the three ligands from condensation reaction of 3,4-dihydroxybenzaldehyde (L'H2)
Ting Xie et al.
PeerJ, 7, e7690-e7690 (2019-10-03)
Lecanicillium lecanii is an entomopathogenic fungi, which was isolated from insects suffering from disease. Now, it is an effective bio-control resource that can control agricultural pests such as whitefly and aphids. There are many studies on the control of various
Ryohei Kono et al.
Acta histochemica et cytochemica, 47(3), 103-112 (2014-10-17)
Granulosa cells form ovarian follicles and play important roles in the growth and maturation of oocytes. The protection of granulosa cells from cellular injury caused by oxidative stress is an effective therapy for female infertility. We here investigated an effective
Nicole L Ritzert et al.
Faraday discussions, 172, 27-45 (2014-11-27)
Over the past decade, there has been a great deal of interest in graphene with regards to its electrochemical behavior. Previous studies have focused on understanding fundamental processes such as charge transfer and molecular transport at the graphene-electrolyte interface as
Tatjana D Savić et al.
Physical chemistry chemical physics : PCCP, 16(38), 20796-20805 (2014-08-29)
The surface modification of nanocrystalline TiO2 particles (45 Å) with catecholate-type ligands having different electron donating/electron withdrawing substituent groups, specifically 3-methylcatechol, 4-methylcatechol, 3-methoxycatechol, 3,4-dihydroxybenzaldehyde and 4-nitrocatechol, was found to alter the optical properties of nanoparticles in a similar way to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service