Skip to Content
Merck
All Photos(1)

Key Documents

35409

Supelco

β-Zearalenol solution

~10 μg/mL in acetonitrile, analytical standard

Synonym(s):

2,4-Dihydroxy-6-(6β,10-dihydroxy-trans-1-undecenyl)benzoic acid μ-lactone solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O5
CAS Number:
Molecular Weight:
320.38
Beilstein:
3653579
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

~10 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

C[C@H]1CCC[C@@H](O)CCC\C=C\c2cc(O)cc(O)c2C(=O)O1

InChI

1S/C18H24O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,14,19-21H,2,4-6,8-9H2,1H3/b7-3+/t12-,14-/m0/s1

InChI key

FPQFYIAXQDXNOR-PMRAARRBSA-N

Application

β-Zearalenol may be used as an analytical reference standard for the determination of the analyte in traditional medicinal herbs, human and animal urine by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phytoestrogens in traditional medicinal herbs using gas chromatography-mass spectrometry.
Lee SH, et al.
The Journal of Nutritional Biochemistry, 15(8), 452-460 (2004)
Validation of a UHPLC?MS/MS method for quantification of zearalenone, α-zearalenol, β-zearalenol, α-zearalanol, β-zearalanol and zearalanone in human urine.
Belhassen H, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 962(8), 68-74 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service