Skip to Content
Merck
All Photos(1)

Documents

34534

Supelco

Imidacloprid-olefin

PESTANAL®, analytical standard

Synonym(s):

1-[(6-Chloro-3-pyridinyl)methyl]-N-nitro-1H-imidazol-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8ClN5O2
CAS Number:
Molecular Weight:
253.65
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

ClC1=NC=C(CN2C(N[N+]([O-])=O)=NC=C2)C=C1

InChI

1S/C9H8ClN5O2/c10-8-2-1-7(5-12-8)6-14-4-3-11-9(14)13-15(16)17/h1-5H,6H2,(H,11,13)

InChI key

TYLCDJYHUVCRBH-UHFFFAOYSA-N

General description

Imidacloprid-olefin is a metabolite of the insecticide, imidacloprid.

Find more information here: Neonicotinoids

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Persistence and metabolism of imidacloprid in different soils of West Bengal.
Sarkar AM, et al.
Pest Management Science, 57(7), 598-602 (2001)
Natural Beekeeping: Organic Approaches to Modern Apiculture, 2nd Edition (2013)
Maura J Hall et al.
Molecules (Basel, Switzerland), 25(12) (2020-06-18)
Consistent with the large-scale use of pesticide seed treatments in U.S. field crop production, there has been an increased use of neonicotinoid-treated corn and soybean seed over the past decade. Neonicotinoids can move downwind to adjacent off-field pollinator habitats in
Margaret L Eng et al.
The Science of the total environment, 760, 143409-143409 (2020-11-22)
Neonicotinoids are the most widely used insecticides globally, but their rapid metabolism in vertebrates makes diagnosing wildlife exposure challenging. More detailed information on the pattern of imidacloprid metabolites over time could be used to better approximate the timing and level

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Analysis of banned neonicotinoid insecticides from dandelion blossoms using QuEChERS and LC-MS.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service