Skip to Content
Merck
All Photos(1)

Key Documents

34091

Supelco

Flubendazol

VETRANAL®, analytical standard

Synonym(s):

Flumoxanal, Methyl [5-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12FN3O3
CAS Number:
Molecular Weight:
313.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccc(F)cc3

InChI

1S/C16H12FN3O3/c1-23-16(22)20-15-18-12-7-4-10(8-13(12)19-15)14(21)9-2-5-11(17)6-3-9/h2-8H,1H3,(H2,18,19,20,22)

InChI key

CPEUVMUXAHMANV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flubendazol is a broad spectrum anthelmintic, which belongs to the class of compounds known as benzimidazoles. It can be widely used in veterinary and human medicine. It mainly finds applications in deworming poultry, swine, dogs and cats.
Flubendazol is an injectable benzimidazole drug; though it is poorly absorbed it is quite effective in the treatment of trycocephalus, ascaris and oxyuriasis in both children and adults.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
VETRANAL® analytical standard can be used in the liquid chromatography coupled to mass spectrometry (LC-MS/MS) procedure, performed for the analysis of macrocyclic lactones in milk.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriel Rübensam et al.
Analytica chimica acta, 705(1-2), 24-29 (2011-10-04)
In this work a method is proposed and demonstrated for the analysis of the macrocyclic lactones abamectin, doramectin, eprinomectin, ivermectin and moxidectin in bovine milk by liquid chromatography coupled to mass spectrometry (LC-MS/MS) and liquid chromatography with fluorescence detection (LC-FL).
Paul A Spagnuolo et al.
Blood, 115(23), 4824-4833 (2010-03-30)
On-patent and off-patent drugs with previously unrecognized anticancer activity could be rapidly repurposed for this new indication given their prior toxicity testing. To identify such compounds, we conducted chemical screens and identified the antihelmintic flubendazole. Flubendazole induced cell death in
H Bártíková et al.
Journal of veterinary pharmacology and therapeutics, 33(1), 56-62 (2010-05-07)
The aim of this project was to study the influence of haemonchosis, a common parasitic infection of small ruminants caused by Haemonchus contortus, on the activity of biotransformation enzymes and on in vitro flubendazole (FLU) biotransformation in liver and small
Integrated pharmacological assessment of flubendazole potential for use in sheep: disposition kinetics, liver metabolism and parasite diffusion ability
Moreno.L, et al.
Journal of Veterinary Pharmacology and Therapeutics, 27, 299-308 (2004)
S Squires et al.
Veterinary parasitology, 185(2-4), 352-354 (2011-10-26)
The efficacy of a commercially available flubendazole-based product and a commercially available herbal product were compared against three species of helminth parasites of chickens: Ascaridia galli, Heterakis gallinarum and Capillaria spp. A total of 48 naturally infected chickens were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service