Skip to Content
Merck
All Photos(1)

Documents

33972

Supelco

3,5,6-Trichloro-2-pyridinol

PESTANAL®, analytical standard

Synonym(s):

2,3,5-Trichloro-6-hydroxypyridine, 3,5,6-Trichloro-2-pyridone, TCP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2Cl3NO
CAS Number:
Molecular Weight:
198.43
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1nc(Cl)c(Cl)cc1Cl

InChI

1S/C5H2Cl3NO/c6-2-1-3(7)5(10)9-4(2)8/h1H,(H,9,10)

InChI key

WCYYAQFQZQEUEN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5,6-Trichloro-2-pyridinol (TCP) is an organophosphorous insecticide, which can be obtained as a major degradation product of chlorpyrifos (CP), via treatment with the degrading bacteria Enterobacter B-14.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Castilho et al.
Journal of the mechanical behavior of biomedical materials, 65, 344-355 (2016-09-16)
Tibial tuberosity advancement (TTA) is a promising method for the treatment of cruciate ligament rupture in dogs that usually implies the implantation of a titanium cage as bone implant. This cage is non-biodegradable and fails in providing adequate implant-bone tissue
Development of an enzyme-linked immunosorbent assay for 3, 5, 6-trichloro-2-pyridinol. 2. Assay optimization and application to environmental water samples
Manclus.JJ and Montoya A
Journal of Agricultural and Food Chemistry, 44, 3710-3716 (1996)
Zuyan Lu et al.
Biofabrication, 11(4), 045012-045012 (2019-07-18)
Supplying oxygen to inner areas of cell constructs to support cell proliferation and metabolism is a major challenge in tissue engineering involving stem cells. Developing devices that incorporate oxygen release materials to increase the availability of the localized oxygen supply
Biodegradation of chlorpyrifos and 3, 5, 6-trichloro-2-pyridinol by a newly isolated Paracoccus sp. strain TRP
Xu G, et al.
International Biodeterioration & Biodegradation, 62, 51-56 (2008)
Samet Tosun et al.
Scanning, 38(6), 619-624 (2016-04-12)
The purpose of this study is to evaluate the dentinal tubule occlusion potential and penetration of Clinpro® White Varnish (5% sodium fluoride + tri-calcium phosphate) in the presence or absence of Nd:YAG laser. Seventy-five dentin samples collected from 38 freshly extracted human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service