Skip to Content
Merck
All Photos(1)

Key Documents

33387

Supelco

Aldicarb-sulfone

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O4S
CAS Number:
Molecular Weight:
222.26
Beilstein:
2215242
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water (Karl Fischer)

mp

132-135 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(ON=CC(C)(C)S(C)(=O)=O)=O

InChI

1S/C7H14N2O4S/c1-7(2,14(4,11)12)5-9-13-6(10)8-3/h5H,1-4H3,(H,8,10)

InChI key

YRRKLBAKDXSTNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aldicarb-sulfone is a metabolite of the insecticide aldicarb used to control insects, mites and nematodes in agricultural crops.

Application

Aldicarb-sulfone may be used as a reference standard for the determination of the analyte in tobacco using high-performance liquid chromatography with dual post-column reaction and fluorescence detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transport of organic contaminants in groundwater: gas chromatography model to forecast the significance as applied to aldicarb sulfone residues.
J P Mestres et al.
Archives of environmental contamination and toxicology, 16(6), 649-656 (1987-11-01)
Y Picó et al.
Food additives and contaminants, 7 Suppl 1, S29-S34 (1990-01-01)
Aldicarb was applied to the soil of Late Valencia orange field plots. A comparative study between four analytical methods is reported, the NPD-gas chromatography method being the best choice. The work also reports the study of accumulation and persistence of
X L Dai
Zhonghua yu fang yi xue za zhi [Chinese journal of preventive medicine], 25(3), 136-138 (1991-05-01)
A method is described for the analysis of aldicarb and its metabolites in urine by GC/FPD. The sample was concentrated with activated charcoal-Florisil column chromatography, eluted by dichloromethane-acetone (1:1v/v). The aldicarb, aldicarb sulfoxide in the eluted solution were oxidized by
A T Lemley et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 18(2), 189-206 (1983-01-01)
The kinetics of degradation of aqueous solutions of aldicarb, aldicarb sulfoxide and aldicarb sul fone by base hydrolysis were investigated. Pseudo first order rate constants of 37 micrograms/l solutions were determined at different hydroxide concentrations by acid-base titration. Second order
L Fava et al.
Pest management science, 57(12), 1135-1141 (2002-01-23)
The metabolites of pesticides can contaminate groundwater and pose a risk to human health when this water is used for drinking. This paper reports the results of a laboratory study on aldicarb and its main metabolites, aldicarb sulfone and aldicarb

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service