Skip to Content
Merck
All Photos(1)

Documents

32766

Supelco

Tembotrione

PESTANAL®, analytical standard

Synonym(s):

2-{2-Chloro-4-(methylsulfonyl)-3-[(2,2,2-trifluoroethoxy)methyl]benzoyl}-1,3-cyclohexanedione, 2-{2-Chloro-4-mesyl-3-[(2,2,2-trifluoroethoxy)methyl]benzoyl}cyclohexane-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16ClF3O6S
CAS Number:
Molecular Weight:
440.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CS(=O)(=O)c1ccc(c(Cl)c1COCC(F)(F)F)C(=O)C2C(=O)CCCC2=O

InChI

1S/C17H16ClF3O6S/c1-28(25,26)13-6-5-9(15(18)10(13)7-27-8-17(19,20)21)16(24)14-11(22)3-2-4-12(14)23/h5-6,14H,2-4,7-8H2,1H3

InChI key

IUQAXCIUEPFPSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tembotrione is a member of β-triketone family of herbicides that is utilized for grass and broadleaf weed control during early to mid-post-emergence of field corn.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tembotrione may be used as a reference standard for the determination of tembotrione in tomato by quick, easy, cheap, effective, rugged and safe (QuEchERS) extraction method and dispersive liquid-liquid microextraction (DLLME) followed by analysis using high performance liquid chromatography (HPLC) with diode array detection (DAD), in honeybees by modified QuEchERS extraction followed by analysis using LC and gas chromatography (GC) combined with tandem mass spectrometry (MS/MS), in food samples by ultra-high performance liquid chromatography (UHPLC) and quadrupole time-of-flight mass spectrometry (QTOFMS) with electrospray ionization and in plant tissues as well as water samples by DLLME, matrix solid-phase dispersion (MSPD), and solid-phase extraction (SPE) combined with HPLC-DAD.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Eyes,Liver,Kidney

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vilena Kašuba et al.
Environmental science and pollution research international, 27(2), 1800-1807 (2019-11-24)
DNA damage in the liver and kidney cells of adult male Wistar rats was studied using the comet assay after a 28-day oral administration of tembotrione at doses of 0.0007, 0.0013 and 0.7 mg/kg b.w./day [AOEL (acceptable operator exposure level)
Screening of over 600 pesticides, veterinary drugs, food-packaging contaminants, mycotoxins, and other chemicals in food by ultra-high performance liquid chromatography quadrupole time-of-flight mass spectrometry (UHPLC-QTOFMS).
Perez-Ortega P, et al.
Food Analytical Methods, 10(5), 1216-1244 (2017)
Extraction procedures for the study of phytotoxicity and degradation processes of selected triketones in a water ecosystem.
Barchanska H, et al.
Environmental Science and Pollution Research International, 21(6), 4751-4758 (2014)
Monopersulfate photocatalysis under 365 nm radiation. Direct oxidation and monopersulfate promoted photocatalysis of the herbicide tembotrione.
Solis RR, et al.
Journal of Environmental Management, 181, 385-394 (2016)
Anita Küpper et al.
Pest management science, 74(10), 2325-2334 (2017-11-07)
Resistance to the 4-hydroxyphenylpyruvate dioxygenase (HPPD)-inhibiting herbicide tembotrione in an Amaranthus palmeri population from Nebraska (NER) has previously been confirmed to be attributable to enhanced metabolism. The objective of this study was to identify and quantify the metabolites formed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service