Skip to Content
Merck
All Photos(1)

Documents

27506

Sigma-Aldrich

L-(+)-Tartaric acid

puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder

Synonym(s):

(2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725147
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

vapor density

5.18 (vs air)

Quality Level

grade

puriss.

Assay

99.7-100.5% (calc. to the dried substance)

form

powder

optical activity

[α]20/D +12.0 to +12.8°, c = 20% in H2O

autoignition temp.

797 °F

quality

meets analytical specification of Ph. Eur., NF

impurities

organic volatile impurities, complies (GC)
residual solvents, complies
≤0.001% heavy metals (as Pb)

ign. residue

≤0.05% (as SO4)

loss

≤0.2% loss on drying, 105 °C

mp

168-170 °C
170-172 °C (lit.)

solubility

water: soluble 150 g/L at 20 °C

anion traces

chloride (Cl-): ≤100 mg/kg
oxalate (C2O42-): in accordance
oxalate (as oxalic acid): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤200 mg/kg
Cu: ≤10 mg/kg
Hg: ≤1 mg/kg
Pb: ≤2 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

application(s)

pharmaceutical (small molecule)

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Cole et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 35(2), 203-206 (1987-02-01)
Tartrate-resistant acid phosphatase (TRAP) has been proposed as a cytochemical marker for osteoclasts. We have developed an improved technique for the localization of TRAP in rat and mouse bone and cartilage. This procedure employs JB-4 plastic as the embedding medium
M Alsina et al.
Blood, 87(4), 1495-1501 (1996-02-15)
Osteolytic bone destruction and its complications, bone pain, pathologic fractures, and hypercalcemia, are a major source of morbidity and mortality in patients with multiple myeloma. The bone destruction in multiple myeloma is due to increased osteoclast (OCL) activity and decreased
Mrinal Kanti Bain et al.
Carbohydrate polymers, 91(2), 529-536 (2012-11-06)
Gelation temperature of MC was reduced from 59°C to 54°C with the addition of 10% PEG. Sodium tartrate (NaT) and sodium citrate (NaC) were added to the MC-PEG solution to further reduce the gelation temperature close to physiological temperature. Different
Mark D Eddleston et al.
Chemical communications (Cambridge, England), 48(92), 11340-11342 (2012-10-18)
The formation of diastereomeric cocrystals of malic acid and tartaric acid was investigated by liquid-assisted grinding in the solid state. We demonstrate that racemic malic acid can be converted into two distinct diastereomeric cocrystal phases by grinding with a single
J B Olivato et al.
Carbohydrate polymers, 92(2), 1705-1710 (2013-02-13)
Tartaric acid (TA), a dicarboxylic acid, can act as a compatibiliser in starch/polyester blends. A mixture design was proposed to evaluate the effect of TA on the properties of starch/poly (butylene adipate co-terephthalate) (PBAT) blown films plasticised with glycerol. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service