Recommended Products
Quality Level
Assay
99.5% trace metals basis
form
powder
reaction suitability
reagent type: oxidant
density
1.95 g/mL at 25 °C (lit.)
SMILES string
N.OCl(=O)(=O)=O
InChI
1S/ClHO4.H3N/c2-1(3,4)5;/h(H,2,3,4,5);1H3
InChI key
HHEFNVCDPLQQTP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Ammonium perchlorate is widely used as an oxidizing agent. The orthorhombic crystals of NH4ClO4 converts to cubic crystal system at temperatures higher than 240°C. CuO nanorods promotes the thermal degradation of ammonium perchlorate.
Application
Ammonium perchlorate (NH4ClO4) may be employed for the purification of as-made “pure” mesoporous silicas via oxidation.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Ox. Sol. 1 - STOT RE 2
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of CuO nanorods and their catalytic activity in the thermal decomposition of ammonium perchlorate
J. Alloy Compounds, 464(1), 532-536 (2008)
Mesoporous silicas prepared by ammonium perchlorate oxidation and theirs application in the selective adsorption of high explosives
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 168, 46-50 (2013)
Biomedical chromatography : BMC, 24(3), 318-323 (2009-07-28)
HPLC columns packed with 3 microm particle size HPLC Technology Techsphere SCX (propylsulfonic acid-modified) silica offer considerable advantages over 5 microm SCX packings in the analysis of basic drugs using 100% methanol eluents containing an ionic modifier such as ammonium
Toxicology, 228(1), 58-65 (2006-10-03)
The U.S. EPA Endocrine Disruptor Screening Program (EDSP) Tier 1 male pubertal protocol was designed as a screen to detect endocrine-disrupting chemicals which may alter reproductive development or thyroid function. One purpose of this in vivo screening protocol is to
Journal of environmental quality, 37(2), 308-317 (2008-02-13)
Within Canada, it has been recognized in the last decade that military training activities may have impacts on the environmental quality of training ranges. However, impacts of activities specific to Air Force Bases have not yet been intensely documented. A
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service