Skip to Content
Merck
All Photos(1)

Key Documents

177539

Sigma-Aldrich

Syringaldazine

98%, powder or crystals

Synonym(s):

4-Hydroxy-3,5-dimethoxybenzaldehyde azine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H2(OCH3)2CH=N-]2
CAS Number:
Molecular Weight:
360.36
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

Syringaldazine, 98%

Quality Level

Assay

98%

form

powder or crystals

mp

209-210 °C (lit.)

solubility

DMF: 5%, clear to hazy

storage temp.

2-8°C

SMILES string

COc1cc(\C=N\N=C\c2cc(OC)c(O)c(OC)c2)cc(OC)c1O

InChI

1S/C18H20N2O6/c1-23-13-5-11(6-14(24-2)17(13)21)9-19-20-10-12-7-15(25-3)18(22)16(8-12)26-4/h5-10,21-22H,1-4H3/b19-9+,20-10+

InChI key

YARKTHNUMGKMGS-LQGKIZFRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Syringaldazine has been used as a substrate to measure the enzymatic activity of recombinant laccase.

Biochem/physiol Actions

Syringaldazine (4-Hydroxy-3, 5-dimethoxybenzaldehyde azine) is a nonautooxidizable laccase-specific compound. It acts as a substrate for laccase and peroxidase in the presence of hydrogen peroxide (H2O2). Syringaldazine also acts as an electron donor in the prostaglandin H synthase (PGHS) reaction.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Bao et al.
Science (New York, N.Y.), 260(5108), 672-674 (1993-04-30)
Peroxidase has been thought to be the only enzyme that oxidizes monolignol precursors to initiate lignin formation in plants. A laccase was purified from cell walls of differentiating xylem of loblolly pine and shown to coincide in time and place
Y A Kuznetsova et al.
Applied biochemistry and biotechnology, 61(1-2), 205-209 (1996-10-01)
The prostaglandin H synthase (PGHS) reaction has been studied with syringaldazine as the electron donor. Kinetic parameters of the reaction agreed with those for commonly used electron donors. The sensitivity of the PGHS activity detection in the presence of syringaldazine
Use of syringaldazine for detection of laccase in sporophores of wood rotting fungi.
J M Harkin et al.
Mycologia, 66(3), 469-476 (1974-05-01)
S Molinari
Journal of nematology, 23(2), 254-258 (1991-04-01)
Isoperoxidases were detected in resistant Rossol and susceptible Roma VF tomato roots uninfected and infected by Meloidogyne incognita. Syringaldazine, guaiacol, p-phenylenediamine-pyrocatechol (PPD-PC), and indoleacetic acid (IAA) were used as substrates, and the corresponding peroxidative activities were detected either in cytoplasmic
C L Wabner et al.
The Journal of urology, 149(6), 1405-1408 (1993-06-01)
The value of orange juice consumption in kidney stone prevention was examined in 8 healthy men and 3 men with documented hypocitraturic nephrolithiasis. They underwent 3 phases of a metabolic study, a placebo phase and 2 treatment phases in which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service